Skip to Content
Merck
All Photos(1)

Key Documents

Y0001511

Glibenclamide for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glybenclamide, 5-Chloro-N-[4-(cyclohexylureidosulfonyl)phenethyl]-2-methoxybenzamide, Glyburide, N-p-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonyl-N′-cyclohexylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28ClN3O5S
CAS Number:
Molecular Weight:
494.00
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glibenclamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3

InChI

1S/C23H28ClN3O5S/c1-32-21-12-9-17(24)15-20(21)22(28)25-14-13-16-7-10-19(11-8-16)33(30,31)27-23(29)26-18-5-3-2-4-6-18/h7-12,15,18H,2-6,13-14H2,1H3,(H,25,28)(H2,26,27,29)

InChI key

ZNNLBTZKUZBEKO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glibenclamide for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Faiyaz Shakeel et al.
Pharmaceutical development and technology, 19(6), 702-707 (2013-08-13)
Temperature-dependent solubility data of glibenclamide (GBN) in various ethanol-water mixtures is not reported in literature so far. Therefore, the aim of this study was to determine the mole fraction solubility of GBN in various ethanol-water mixtures at the temperature range
J M Feldman
The American journal of medicine, 79(3B), 102-108 (1985-09-20)
After one year on the American market, glyburide, a second-generation sulfonylurea, seems well accepted by physicians. If the hyperglycemia of patients with type II diabetes mellitus is not corrected by diet and exercise, glyburide can be used as adjunctive therapy.
N M O'Meara et al.
The American journal of medicine, 89(2A), 11S-16S (1990-08-20)
Since the introduction of glyburide in 1984, many studies have evaluated the effects of this oral hypoglycemic agent on beta cell function in patients with non-insulin-dependent diabetes mellitus. The early studies, which were performed in patients receiving concomitant insulin therapy
George E Dailey
Expert opinion on pharmacotherapy, 4(8), 1417-1430 (2003-07-25)
Oral antidiabetic combination therapy is a proven means of establishing glycaemic control in the hyperglycaemic, Type 2 diabetic patient, but co-administering two oral antidiabetic agents separately may hinder compliance with therapy. A new single-tablet of glyburide/metformin combination therapy (Glucovance), Bristol-Myers
Thomas R Moore
Diabetes care, 30 Suppl 2, S209-S213 (2008-02-27)
The clinical experience with glyburide treatment of GDM has moved ahead of the science. A single randomized controlled trial of glyburide versus insulin indicates that glyburide treatment can provide a relatively safe alternative to insulin therapy. Subsequent retrospective trials have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service