Skip to Content
Merck
All Photos(1)

Key Documents

Y0001493

Furosemide for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Furosemide, 4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfonyl)-4-chloro-2-([2-furanylmethyl]amino)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

furosemide

manufacturer/tradename

EDQM

mp

220 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NS(=O)(=O)c1cc(C(O)=O)c(NCc2ccco2)cc1Cl

InChI

1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)

InChI key

ZZUFCTLCJUWOSV-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Furosemide for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Inhibits ion co-transport in the kidney.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam D DeVore et al.
European journal of heart failure, 17(3), 340-346 (2015-01-27)
Loop diuretics are a cornerstone of heart failure (HF) treatment, but data regarding diuretic dose adjustments after a HF hospitalization and the association with subsequent outcomes are limited. This study was therefore conducted to determine these factors. We analysed data
Christianne L Roumie et al.
Stroke, 46(2), 465-470 (2015-01-01)
We examined blood pressure 1 year after stroke discharge and its association with treatment intensification. We examined the systolic blood pressure (SBP) stratified by discharge SBP (≤140, 141-160, or >160 mm Hg) among a national cohort of Veterans discharged after
K M Ho et al.
Anaesthesia, 65(3), 283-293 (2010-01-21)
Furosemide, a potent loop diuretic, is frequently used in different stages of acute kidney injury, but its clinical roles remain uncertain. This review summarises the pharmacology of furosemide, its potential uses and side effects, and the evidence of its efficacy.
Phillip J Newton et al.
Journal of pain and symptom management, 36(4), 424-441 (2008-05-13)
Dyspnea is a common and distressing symptom associated with multiple chronic illnesses and high levels of burden for individuals, their families and health care systems. The subjective nature dyspnea and a poor understanding of pathophysiological mechanisms challenge the clinician in
Kurt A Wargo et al.
The Annals of pharmacotherapy, 43(11), 1836-1847 (2009-10-22)
To review the literature regarding the pharmacokinetic profiles, comparative safety and efficacy, and comparative costs of loop diuretics to evaluate the current clinical usefulness of furosemide. A search of MEDLINE (1966-June 2009) was conducted using the terms furosemide, torsemide, bumetanide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service