Skip to Content
Merck
All Photos(1)

Key Documents

Y0001474

Lauric acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dodecanoic acid, Lauric acid, NSC 5026

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COOH
CAS Number:
Molecular Weight:
200.32
Beilstein:
1099477
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lauric acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC(O)=O

InChI

1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)

InChI key

POULHZVOKOAJMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lauric acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Substrate for CYP 4A11

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

348.8 °F - closed cup

Flash Point(C)

176 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Mesman et al.
Bipolar disorders, 17(1), 39-49 (2014-07-22)
There is increasing evidence that both immune and neurochemical alterations are involved in the pathogenesis of bipolar disorder; however, their precise role remains unclear. In this study, we aimed to evaluate neuro-immune changes in a prospective study on children of
Malin Holzmann et al.
European journal of obstetrics, gynecology, and reproductive biology, 184, 97-102 (2014-12-09)
To investigate if repeat (≥ 3) fetal scalp blood sampling (FBS) is associated with increased risk of caesarean delivery and worse neonatal outcome than occasional (1-2) FBS. Prospective cohort study of women undergoing intrapartum FBS at Karolinska University Hospital, Sweden.
Avichai Shimoni et al.
Cancer, 121(6), 863-871 (2014-11-13)
Allogeneic stem cell transplantation (SCT) remains the standard treatment for advanced chronic myeloid leukemia (CML) and Philadelphia chromosome-positive acute lymphoblastic leukemia (Ph(+) ALL). Relapsed disease is the major cause of treatment failure, especially when SCT is given in the setting
Meng-Meng Bai et al.
Journal of agricultural and food chemistry, 63(8), 2198-2205 (2015-02-14)
Eucommia ulmoides leaves have been used as a functional food and drink in China. The purpose of this study was to identify the bioactive constituents with soluble epoxide hydrolase (sEH) inhibitory activity and anti-inflammatory properties. Twenty-seven known compounds (1-27) were
M Schmidt et al.
Leukemia, 28(12), 2292-2299 (2014-09-13)
To study clonal evolution in chronic myeloid leukemia (CML), we searched for BCR-ABL-independent gene mutations in both Philadelphia chromosome (Ph)-negative and Ph-positive clones in 29 chronic-phase CML patients by targeted deep sequencing of 25 genes frequently mutated in myeloid disorders.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service