Skip to Content
Merck
All Photos(1)

Key Documents

Y0001410

Sodium fusidate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fusidic acid sodium salt, Fucidin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C31H47O6Na
CAS Number:
Molecular Weight:
538.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sodium fusidate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].C[C@@H]1[C@H](O)CC[C@@]2(C)C1CC[C@@]3(C)C2[C@H](O)CC4\C([C@H](C[C@]34C)OC(C)=O)=C(/CC\C=C(\C)C)C([O-])=O

InChI

1S/C31H48O6.Na/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);/q;+1/p-1/b26-20-;/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-;/m0./s1

InChI key

HJHVQCXHVMGZNC-JCJNLNMISA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium fusidate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Suppresses nitric oxide lysis of pancreatic islet cells. Inhibits protein synthesis in prokaryotes by inhibiting the ribosome-dependent activity of G factor and translocation of peptidyl-tRNA.
Suppresses nitric oxide lysis of pancreatic islet cells; inhibits protein synthesis in prokaryotes.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of fusidic acid and sodium fusidate in pharmaceutical dosage forms by first-derivative ultraviolet spectrophotometry.
Hassan S M, et al.
Analyst, 112(10), 1459-1461 (1987)
Topical antibiotics in the treatment of superficial skin infections in general practice?a comparison of mupirocin with sodium fusidate.
White D G, et al.
The Journal of Infection, 18(3), 221-229 (1989)
Saleem Shaikh et al.
Journal of chromatographic science, 47(2), 178-183 (2009-02-19)
A simple, specific, and precise high-performance liquid chromatographic method is developed and validated for the simultaneous determination of chlorocresol (CC), mometasone furoate (MF), and fusidic acid (FA) in a cream formulation. The isocratic mobile phase consists of 1.5% w/v aqueous
Katharina Wittmann et al.
Tissue engineering. Part A, 21(7-8), 1343-1353 (2015-01-21)
The development of vascularized and functional adipose tissue substitutes is required to improve soft tissue augmentation. In this study, vascularized adipose tissue constructs were generated using uncultured cells from the stromal-vascular fraction (SVF) of adipose tissue as an alternative cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service