Skip to Content
Merck
All Photos(3)

Documents

F0881

Sigma-Aldrich

Fusidic acid sodium salt

≥98% (TLC)

Synonym(s):

Fucidin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C31H47O6Na
CAS Number:
Molecular Weight:
538.69
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

solubility

H2O: soluble

antibiotic activity spectrum

neoplastics

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].C[C@@H]1[C@H](O)CC[C@@]2(C)C1CC[C@@]3(C)C2[C@H](O)CC4\C([C@H](C[C@]34C)OC(C)=O)=C(/CC\C=C(\C)C)C([O-])=O

InChI

1S/C31H48O6.Na/c1-17(2)9-8-10-20(28(35)36)26-22-15-24(34)27-29(5)13-12-23(33)18(3)21(29)11-14-30(27,6)31(22,7)16-25(26)37-19(4)32;/h9,18,21-25,27,33-34H,8,10-16H2,1-7H3,(H,35,36);/q;+1/p-1/b26-20-;/t18-,21-,22-,23+,24+,25-,27-,29-,30-,31-;/m0./s1

InChI key

HJHVQCXHVMGZNC-JCJNLNMISA-M

Looking for similar products? Visit Product Comparison Guide

General description

Fusidic acid is an antibiotic substance, resembling the structure of a steroid. It belongs to the class of fusidanes.

Application

Fusidic acid sodium salt has been used to study the effects of three-drug combinations on the growth rate of a bacterium, wild-type Escherichia coli. It is also used to study the in vitro susceptibility of dog-derived European methicillin-resistant Staphylococcus pseudintermedius and Staphylococcus aureus strains and their methicillin-susceptible counterparts.

Biochem/physiol Actions

Fusidic acid possesses antimicrobial action against skin pathogens. It is highly specific to Staphylococcus aureus. It is therefore used for treating infected traumatic wounds, folliculitis, furunculosis, impetigo, erythrasma and abscesses.
Suppresses nitric oxide lysis of pancreatic islet cells. Inhibits protein synthesis in prokaryotes by inhibiting the ribosome-dependent activity of G factor and translocation of peptidyl-tRNA.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Susceptibility in vitro of canine methicillin-resistant and-susceptible staphylococcal isolates to fusidic acid, chlorhexidine and miconazole: opportunities for topical therapy of canine superficial pyoderma
<BIG>Clark SM, et al. </BIG>
The Journal of Antimicrobial Chemotherapy, 2048-2052 (2015)
Fusidic acid in dermatology
British Journal of Dermatology, 37-40 (1998)
Enhanced identification of synergistic and antagonistic emergent interactions among three or more drugs
<BIG>Tekin E, et al.</BIG>
Journal of the Royal Society, Interface / the Royal Society, 20160332-20160332 (2016)
David J F Ramrath et al.
Nature, 485(7399), 526-529 (2012-05-25)
Bacterial ribosomes stalled at the 3' end of malfunctioning messenger RNAs can be rescued by transfer-messenger RNA (tmRNA)-mediated trans-translation. The SmpB protein forms a complex with the tmRNA, and the transfer-RNA-like domain (TLD) of the tmRNA then enters the A
R Di Marco et al.
Journal of autoimmunity, 13(2), 187-195 (1999-09-10)
The immunomodulating antibiotic drug fusidic acid and its sodium salt sodium fusidate (fusidin) ameliorate several organ-specific immunoinflammatory diseases. Because preliminary observations suggest that fusidin may also exert a beneficial effect in Guillain-Barré syndrome (GBS), here we have studied the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service