Skip to Content
Merck
All Photos(1)

Key Documents

Y0001247

Tropicamide impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tropic acid, 2-Phenylhydracrylic acid, 3-Hydroxy-2-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH2OH)CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
2209199
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tropicamide

manufacturer/tradename

EDQM

mp

116-118 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OCC(C(O)=O)c1ccccc1

InChI

1S/C9H10O3/c10-6-8(9(11)12)7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)

InChI key

JACRWUWPXAESPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tropicamide impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dima Albals et al.
Electrophoresis, 35(19), 2807-2818 (2014-07-02)
A generic chiral separation strategy for the analysis of acidic compounds in CEC is proposed in completion of an earlier defined strategy for nonacidic compounds. The screening step of this strategy uses a 45 mM ammonium formate (pH 2.9)/ACN (35/65
Jadwiga Lyczko et al.
Journal of mass spectrometry : JMS, 50(3), 463-469 (2015-03-25)
Common calibration standards for mass spectrometry can be a source of many problems including instrument contamination, ionization suppression and formation of unidentified ions during subsequent analysis. In this article, we present a new approach for the calibration of mass analyzers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service