Skip to Content
Merck
All Photos(1)

Key Documents

Y0001459

Triamcinolone acetonide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Triamcinolone acetonide, 9α-Fluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide, 9α-Fluoro-16α-hydroxyprednisolone 16α,17α-acetonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

triamcinolone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@H]5OC(C)(C)O[C@@]45C(=O)CO

InChI

1S/C24H31FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-9,15-17,19,26,28H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1

InChI key

YNDXUCZADRHECN-JNQJZLCISA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triamcinolone acetonide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mazen Abu-Mugheisib et al.
Current pharmaceutical design, 18(29), 4564-4569 (2012-05-23)
Multiple sclerosis (MS) as a chronic disease presents a vast variability of neurological symptoms. Spasticity is one of the most common symptoms. The principle indication to treat spasticity is disability or handicap of capabilities. All pharmacological methods are attributed to
Daniele Veritti et al.
Expert opinion on drug safety, 11(2), 331-340 (2011-11-10)
Triamcinolone acetonide (TA) is a steroidal drug that has been widely administered intravitreally for retinal and choroidal conditions. Safety of steroidal products for intraocular use is essential because of their risk of ocular adverse events. This review comprehensively discusses the
Hui-Ping Qi et al.
Current eye research, 37(12), 1136-1147 (2012-07-17)
To compare the efficacy of intravitreal (IV) triamcinolone acetonide (IVTA) versus subtenon (ST) triamcinolone acetonide (STTA) injection for the treatment of diabetic macular edema (DME). Searches for randomized clinical trials published between 1 January 1950 and 15 March 2011 were
Rafi Fredman et al.
Burns : journal of the International Society for Burn Injuries, 39(4), 549-557 (2012-10-25)
Intralesional triamcinolone acetonide (TAC) is a well-established treatment for keloids and hypertrophic scars. The present text provides a systematic review of all previously reported cases of Cushing's syndrome resulting from intralesional TAC in an effort to discover whether an association
Jost B Jonas
Ophthalmic research, 38(4), 218-245 (2006-06-10)
Based on experimental studies and clinical observations by Robert Machemer, Gholam Peyman and others, the vitreous cavity has increasingly been used as a reservoir of drugs for the direct treatment of intraocular diseases. The most widely injected drug so far

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service