Skip to Content
Merck
All Photos(1)

Key Documents

Y0001163

Ursodeoxycholic acid for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ursodeoxycholic acid, 3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid-3α,7β-diol, 7β-Hydroxylithocholic acid, UDCS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Beilstein:
3219888
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ursodeoxycholic acid

manufacturer/tradename

EDQM

mp

203-204 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1

InChI key

RUDATBOHQWOJDD-UZVSRGJWSA-N

Gene Information

human ... NR1H4(9971)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ursodeoxycholic acid for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John E Eaton et al.
Gastroenterology, 145(3), 521-536 (2013-07-06)
Primary sclerosing cholangitis (PSC), first described in the mid-1850s, is a complex liver disease that is heterogeneous in its presentation. PSC is characterized by chronic cholestasis associated with chronic inflammation of the biliary epithelium, resulting in multifocal bile duct strictures
Li-Na Zhang et al.
Hepatology (Baltimore, Md.), 58(1), 264-272 (2013-02-15)
The biochemical response to ursodeoxycholic acid (UDCA) in primary biliary cirrhosis is a strong predictor of long-term outcome and thus facilitates the rapid identification of patients needing new therapeutic approaches. Numerous criteria for predicting outcome of treatment have been studied
Marco Carbone et al.
Gastroenterology, 144(3), 560-569 (2012-12-19)
BACKGROUND, & AIMS: Studies of primary biliary cirrhosis (PBC) phenotypes largely have been performed using small and selected populations. Study size has precluded investigation of important disease subgroups, such as men and young patients. We used a national patient cohort
Heather Mortiboys et al.
Brain : a journal of neurology, 136(Pt 10), 3038-3050 (2013-09-04)
Previous drug screens aiming to identify disease-modifying compounds for Parkinson's disease have typically been based on toxin-induced in vitro and in vivo models of this neurodegenerative condition. All these compounds have failed to have a reliable disease-modifying effect in subsequent
L Serfaty et al.
Gastroenterologie clinique et biologique, 34(10), 516-522 (2010-07-09)
Colorectal cancer is respectively the third and second most common cancer among men and women in France. Interest in chemoprevention for colorectal cancer has increased over the last two decades. Beside non-steroidal anti-inflammatory drugs, ursodeoxycholic acid (UDCA) may have chemopreventive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service