Skip to Content
Merck
All Photos(1)

Key Documents

Y0000761

Pholcodine

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N2O4
CAS Number:
Molecular Weight:
398.50
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pholcodine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N6(CCOCC6)CCOc1c2c3c(cc1)C[C@H]4N(CC[C@]53[C@@H](O2)[C@H](C=CC54)O)C

InChI

1S/C23H30N2O4/c1-24-7-6-23-16-3-4-18(26)22(23)29-21-19(5-2-15(20(21)23)14-17(16)24)28-13-10-25-8-11-27-12-9-25/h2-5,16-18,22,26H,6-14H2,1H3/t16?,17-,18+,22+,23+/m1/s1

InChI key

GPFAJKDEDBRFOS-SLITUTQKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pholcodine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivien Tong et al.
Health expectations : an international journal of public participation in health care and health policy, 19(4), 948-961 (2015-08-01)
Over-the-counter (OTC) labels help support safe medication use by consumers. In 2012, the Australian Therapeutic Goods Administration (TGA) released a consultation paper proposing OTC label standardization to improve label quality via implementation of the Medicine Information Box (MIB) label. However
Philip H Li et al.
The journal of allergy and clinical immunology. In practice, 5(6), 1601-1606 (2017-05-30)
True IgE-mediated hypersensitivity to opioids is rare and many reactions are due to direct mast cell degranulation. Opioid drug provocation testing (DPT) is the gold standard for diagnosis but is underutilized. The objective of this study was to evaluate the
Pholcodine in cough medicines and IgE-sensitization in the EU: an urgent task.
E Florvaag et al.
Allergy, 67(5), 581-582 (2012-03-03)
Paul M Mertes et al.
Current opinion in allergy and clinical immunology, 12(4), 361-368 (2012-07-07)
Immediate hypersensitivity reactions are an important cause for mortality and morbidity in anesthesia. The present review considers reports covering epidemiology, diagnosis, and treatment of these reactions. Immediate hypersensitivity reactions are largely under-reported, adult women being at significantly higher risk than
C Meadway et al.
Forensic science international, 127(1-2), 131-135 (2002-07-06)
Although the cross reactivity of pholcodine with opiate immunoassays has been well documented there is little published information the potential for pholcodine interference with chromatographic analyses. Wilson and Smith [Ann. Clin. Biochem. 36 (1999) 592] recently described the 'misidentification' of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service