Skip to Content
Merck
All Photos(1)

Key Documents

Y0000641

Azithromycin for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Azithromycin dihydrate, 1-Oxa-6-azacyclopentadecan-15-one, 13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-alpha-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10- trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-beta-D-xylohexopyranosyl]oxy]-, [2R-(2R, 3S, 4R, 5R, 8R, 10R, 11R, 12S, 13S, 14R)]- dihydrate, N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A, CP-62993

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H72N2O12 · 2H2O
CAS Number:
Molecular Weight:
785.02
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

azithromycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C38H72N2O12.2H2O/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28;;/h20-33,35,41-43,45-46H,15-19H2,1-14H3;2*1H2/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-;;/m1../s1

InChI key

SRMPHJKQVUDLQE-KUJJYQHYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Azithromycin for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Azithromycin dihydrate is a macrolide antibiotic with anti-immunomodulatory/anti-inflammatory properties.
Azithromycin dihydrate is a macrolide antibiotic, azalide subclass. It binds to the 50S subunit of the 70S bacterial ribosomes and inhibits RNA-dependent protein synthesis in bacterial cells. Azithromycin also has anti-immunomodulatory/anti-inflammatory properties, which make it useful in treating cystic fibrosis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alison M Binder et al.
American journal of respiratory and critical care medicine, 188(7), 807-812 (2013-08-10)
Persons with cystic fibrosis (CF) are at high risk of nontuberculous mycobacterial (NTM) infection, with treatment requiring prolonged multidrug regimens that include macrolides. Although macrolides, specifically azithromycin, are used in the management of patients with CF with chronic Pseudomonas, macrolide-resistant
Sheila K West et al.
The Journal of infectious diseases, 210(1), 65-71 (2014-01-22)
Trachoma, caused by repeated infections with ocular Chlamydia trachomatis, is targeted for elimination using multiple annual rounds of mass drug administration (MDA) in endemic communities. Infection rates do not decline as expected in some communities, leading to concerns about azithromycin
Eric M Mortensen et al.
JAMA, 311(21), 2199-2208 (2014-06-04)
Although clinical practice guidelines recommend combination therapy with macrolides, including azithromycin, as first-line therapy for patients hospitalized with pneumonia, recent research suggests that azithromycin may be associated with increased cardiovascular events. To examine the association of azithromycin use with all-cause
Sarah E Burr et al.
Bulletin of the World Health Organization, 92(7), 490-498 (2014-08-12)
To evaluate the effect of repeated mass drug administration (MDA) of azithromycin in the Gambia on the nasopharyngeal carriage of Streptococcus pneumoniae and on the emergence of antibiotic-resistant strains. This study involved villages that participated in a cluster randomized trial
Patricia C Valery et al.
The Lancet. Respiratory medicine, 1(8), 610-620 (2014-01-28)
Indigenous children in high-income countries have a heavy burden of bronchiectasis unrelated to cystic fibrosis. We aimed to establish whether long-term azithromycin reduced pulmonary exacerbations in Indigenous children with non-cystic-fibrosis bronchiectasis or chronic suppurative lung disease. Between Nov 12, 2008

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service