Skip to Content
Merck
All Photos(1)

Documents

Y0000626

Ketorolac trometamol for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ketorolac tris salt, (±)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid tris salt, Toradol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO3 · C4H11NO3
CAS Number:
Molecular Weight:
376.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ketorolac

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(CO)(CO)CO.OC(=O)C1CCn2c1ccc2C(=O)c3ccccc3

InChI

1S/C15H13NO3.C4H11NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13;5-4(1-6,2-7)3-8/h1-7,11H,8-9H2,(H,18,19);6-8H,1-3,5H2

InChI key

BWHLPLXXIDYSNW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ketorolac trometamol for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppina Magni et al.
Anesthesia and analgesia, 116(2), 443-447 (2013-01-11)
Ketorolac tromethamine (ketorolac) is a nonsedating drug with potent analgesic and moderate anti-inflammatory activity, which does not increase the sedation level. The safety of ketorolac with respect to risk of bleeding has been demonstrated in large numbers of patients undergoing
Michel Weber et al.
Acta ophthalmologica, 91(1), e15-e21 (2012-09-14)
To determine whether indomethacin 0.1% eye drops are at least as effective as ketorolac 0.5% eye drops in treating ocular inflammation following cataract surgery. Prospective, multicenter, investigator-masked, parallel-group, randomized, active-controlled clinical trial. Cataract patients were randomized in a 1:1 ratio
Georgios Mylonas et al.
American journal of ophthalmology, 155(2), 253-259 (2012-10-06)
To investigate intraretinal changes after successful cataract surgery in patients with previous pars plana vitrectomy (PPV) and indocyanine green-assisted peeling of epiretinal membrane and internal limiting membrane (ILM). Prospective nonrandomized controlled clinical study. Twenty eyes of 20 patients with cataract
V R Sinha et al.
Expert opinion on drug delivery, 6(9), 961-975 (2009-08-12)
Ketorolac tromethamine (KT) is a non-steroidal anti-inflammatory drug that belongs to the class of heteroaryl acetic acid derivatives. It is a non-selective cyclooxygenase (COX) inhibitor, being marketed in the racemate form. Most of its analgesic and COX inhibitory activity is
Gwen M Grimsby et al.
Mayo Clinic proceedings, 87(11), 1089-1097 (2012-10-13)
To evaluate the efficacy and safety of a novel, continuous intravenous infusion of ketorolac, a powerful nonopioid analgesic, for postoperative pain control. A prospective, double-blind, randomized, placebo-controlled trial of a continuous infusion of ketorolac tromethamine in 1 L of normal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service