Skip to Content
Merck
All Photos(1)

Key Documents

Y0000587

Venlafaxine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(+/-)-1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, Effexor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO2 · HCl
CAS Number:
Molecular Weight:
313.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

venlafaxine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].COc1ccc(cc1)C(CN(C)C)C2(O)CCCCC2

InChI

1S/C17H27NO2.ClH/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14;/h7-10,16,19H,4-6,11-13H2,1-3H3;1H

InChI key

QYRYFNHXARDNFZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Venlafaxine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Venlafaxine is a dual serotonin-noradrenaline reuptake inhibitor that displays Ki values of 82 and 2480 nM respectively. It is a general class antidepressant.
Venlafaxine is an antidepressant. The mechanism of the antidepresant action of venlafaxine in humans is associated with its potentiation of neurotransmitter activity in the CNS. Venlafaxine is a potent inhibitor of neuronal serotonin and norepinephrine reuptake and weak inhibitor of dopamine reuptake. Venlafaxine has no significant activity for muscarinic, histaminergic, or α-1 adrenergic receptors in vitro. Venlafaxine does not possess MAO inhibitor activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Batista et al.
Clinical toxicology (Philadelphia, Pa.), 51(2), 92-95 (2013-01-11)
Venlafaxine is a bicyclic antidepressant that may be associated with severe cardiotoxicity following large overdose. The purpose of this short case series is to present different patterns of venlafaxine-related cardiotoxicity and to discuss the potential mechanisms. Between January 2010 and
Mara Z Vitolins et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(32), 4092-4098 (2013-10-02)
Hot flashes occur in approximately 80% of androgen-deprived men. Few intervention studies have been conducted to relieve hot flashes in men. Eligible androgen-deprived men were randomly assigned to one of four daily regimens (2 × 2 factorial design) for 12
R L Rudolph et al.
Journal of clinical psychopharmacology, 16(3 Suppl 2), 54S-59S (1996-06-01)
The tolerability and safety of venlafaxine hydrochloride, a new serotonin and norepinephrine reuptake inhibitor, are reviewed in this article. The data presented here are based on a pool of 3,082 patients who were treated with this agent during clinical trials.
Yaal Lester et al.
Water research, 47(13), 4349-4356 (2013-06-15)
Wastewater from a pharmaceutical formulation facility (TevaKS, Israel) was treated with a biological activated-sludge system followed by ozonation. The goal was to reduce the concentrations of the drugs carbamazepine (CBZ) and venlafaxine (VLX) before discharging the wastewater to the municipal
Swati Pund et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(1-2), 195-201 (2012-11-20)
Venlafaxine, a dual acting antidepressant is a new therapeutic option for chronic depression. Depression is a common mental disorder associated with the abnormalities in neuronal transport in the brain. Since the nose-to-brain pathway has been indicated for delivering drugs to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service