Skip to Content
Merck
All Photos(1)

Key Documents

V-004

Supelco

Venlafaxine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28ClNO2
CAS Number:
Molecular Weight:
313.86
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.COc1ccc(cc1)C(CN(C)C)C2(O)CCCCC2

InChI

1S/C17H27NO2.ClH/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14;/h7-10,16,19H,4-6,11-13H2,1-3H3;1H

InChI key

QYRYFNHXARDNFZ-UHFFFAOYSA-N

Gene Information

General description

A Certified Snap-N-Spike® Solution suitable for many LC/MS and GC/MS applications from forensic or clinical toxicology analysis to urine drug testing. Also known by the brand name Effexor®, venlafaxine is an SNRI antidepressant approved for the treatment of major depressive and general anxiety disorders.

Application


  • Reliable Dissolved Oxygen Detection: Reliable detection of dissolved oxygen is vital in enzymatic studies and biochemical reactions where oxygen plays a regulatory or reactive role, impacting the kinetics and outcomes of these reactions (Kusanto et al., 2021).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Effexor is a registered trademark of American Home Product Corp.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Depersonalization and derealization syndrome: report on a case study and pharmacological management.
Ana Salgado et al.
Revista brasileira de psiquiatria (Sao Paulo, Brazil : 1999), 34(4), 505-508 (2013-02-23)
Danuta Szkutnik-Fiedler et al.
Pharmacological reports : PR, 64(6), 1350-1358 (2013-02-15)
The aim of this study was to evaluate antidepressant-like effect (Porsolt test), locomotor activity and motor coordination of joint administration of tramadol (TRM) and venlafaxine (VEN) in rats. The tests were performed on male Wistar rats after single and chronic
S Fenli et al.
European review for medical and pharmacological sciences, 17(1), 41-48 (2013-01-19)
Venlafaxine is a new antidepressant that has a chemical structure and neuropharmacologic profile distinct from those of existing antidepressants. The studies about the mechanism of pharmacological action of venlafaxine mostly investigated the effects of venlafaxine on 5-hydroxytryptamine (5-HT), norepinephrine (NE)
M Batista et al.
Clinical toxicology (Philadelphia, Pa.), 51(2), 92-95 (2013-01-11)
Venlafaxine is a bicyclic antidepressant that may be associated with severe cardiotoxicity following large overdose. The purpose of this short case series is to present different patterns of venlafaxine-related cardiotoxicity and to discuss the potential mechanisms. Between January 2010 and
Mandar J Bhandwalkar et al.
AAPS PharmSciTech, 14(1), 101-110 (2012-12-12)
In order to improve the bioavailability of the antidepressant drug, venlafaxine hydrochloride, in situ mucoadhesive thermoreversible gel, was formulated using Lutrol F127 (18%) as a thermo gelling polymer. Mucoadhesion was modulated by trying carbopol 934, PVP K30, HPMC K4M, sodium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service