Skip to Content
Merck
All Photos(1)

Documents

Y0000548

Nandrolone decanoate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

19-Nortestosterone 17-decanoate, 17β-Hydroxy-19-nor-4-androsten-3-one 17-decanoate, 17β-Hydroxy-4-estren-3-one 17-decanoate, 4-Estren-17β-ol-3-one 17-decanoate, Decadurabolin, Nandrolone decanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O3
CAS Number:
Molecular Weight:
428.65
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nandrolone

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C

InChI

1S/C28H44O3/c1-3-4-5-6-7-8-9-10-27(30)31-26-16-15-25-24-13-11-20-19-21(29)12-14-22(20)23(24)17-18-28(25,26)2/h19,22-26H,3-18H2,1-2H3/t22-,23+,24+,25-,26-,28-/m0/s1

InChI key

JKWKMORAXJQQSR-MOPIKTETSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nandrolone decanoate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arnaud Ferry et al.
Endocrinology, 155(12), 4739-4748 (2014-09-13)
The first aim of this study was to examine the role of myofiber androgen receptor (AR) in male mice on muscle performance gain and remodeling-induced muscle mechanical overloading (OVL) that mimics resistance training. The response of OVL in mice in
Martin Elfverson et al.
Neuropharmacology, 61(7), 1172-1181 (2011-01-22)
Studies have shown that anabolic androgenic steroids (AASs) can induce profound changes to mental health. Commonly reported psychiatric side effects among AAS users include aggression, anxiety, depression, drug abuse and cognitive disabilities. In experimental animals, many of these effects have
Gurmeet K S Singh et al.
The Journal of clinical endocrinology and metabolism, 99(7), 2592-2598 (2014-04-02)
Testosterone (T) and nandrolone (N) esters require deep im injections by medical personnel but these often deposit injectate into sc fat so that more convenient sc self-administration may be feasible. To investigate the feasibility and pharmacology of sc injection of
Carolina Almeida do Carmo et al.
Journal of applied toxicology : JAT, 32(10), 810-814 (2011-07-01)
Nandrolone is an androgenic-anabolic steroid (AAS) with diverse medical applications but taken indiscriminately by some to rapidly increase muscle mass. The aim of this study was to evaluate the genotoxic and clastogenic potential of nandrolone (deca-durabolin®) in vivo in different
C M Souza et al.
Journal of medical entomology, 48(1), 111-117 (2011-02-23)
Necrophagous insects are valuable tools for postmortem interval (PMI) estimation or for determining the cause of death. Due to the increase in deaths related to drug abuse, it is crucial to know how these substances affect the development of flies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service