Skip to Content
Merck
All Photos(1)

Key Documents

90151

Supelco

17β-Hydroxy-17-methylandrosta-1,4-dien-3-one

analytical standard

Synonym(s):

1-Dehydro-17α-methyltestosterone, Dianabol, Methandienone, Methandrostenolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
Beilstein:
2056255
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]1.15/D -0.5±0.5° in chloroform

shelf life

limited shelf life, expiry date on the label

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

functional group

ketone

storage temp.

2-8°C

SMILES string

C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C20H28O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h6,9,12,15-17,22H,4-5,7-8,10-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1

InChI key

XWALNWXLMVGSFR-HLXURNFRSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

17β-hydroxy-17-methylandrosta-1,4-dien-3-one is an androgenic anabolic steroid.[1]

Application

17β-hydroxy-17-methylandrosta-1,4-dien-3-one may be used as a reference standard in the determination of 17β-hydroxy-17-methylandrosta-1,4-dien-3-one in urine samples, using by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF-MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative study of matrices for their use in the rapid screening of anabolic steroids by matrix?assisted laser desorption/ionisation time?of?flight mass spectrometry
Galesio.M, et al.
Rapid Communications in Mass Spectrometry, 23, 1783-1791 (2009)
Stephanie Mosler et al.
Archives of toxicology, 86(1), 109-119 (2011-08-06)
There is increasing evidence that the biological activity of myostatin (MSTN), a negative regulator of muscle growth, is affected by training but also anabolic steroids. In this study, we analyzed the effects of the frequently abused anabolic steroid methandienone (Md)
Andy Zöllner et al.
Biological chemistry, 391(1), 119-127 (2009-11-19)
Anabolic-androgenic steroids are some of the most frequently misused drugs in human sports. Recently, a previously unknown urinary metabolite of metandienone, 17beta-hydroxymethyl-17 alpha-methyl-18-norandrosta-1,4,13-trien-3-one (20OH-NorMD), was discovered via LC-MS/MS and GC-MS. This metabolite was reported to be detected in urine samples
Oscar J Pozo et al.
Drug testing and analysis, 1(11-12), 554-567 (2010-04-01)
Anabolic steroids are among the most frequently detected compounds in doping analysis. They are extensively metabolized and therefore an in-depth knowledge about steroid metabolism is needed. In this study, a liquid chromatography tandem mass spectometry (LC-MS/MS) method based on a
Leen Lootens et al.
Clinical chemistry, 55(10), 1783-1793 (2009-08-01)
Adequate detection of designer steroids in the urine of athletes is still a challenge in doping control analysis and requires knowledge of steroid metabolism. In this study we investigated whether uPA(+/+)-SCID mice carrying functional primary human hepatocytes in their liver

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service