Skip to Content
Merck
All Photos(1)

Key Documents

Y0000489

Mycophenolate mofetil

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid 2-(4-morpholinyl)ethyl ester, RS 61443, TM-MMF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31NO7
CAS Number:
Molecular Weight:
433.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

mycophenolate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)CCC(=O)OCCN3CCOCC3

InChI

1S/C23H31NO7/c1-15(5-7-19(25)30-13-10-24-8-11-29-12-9-24)4-6-17-21(26)20-18(14-31-23(20)27)16(2)22(17)28-3/h4,26H,5-14H2,1-3H3/b15-4+

InChI key

RTGDFNSFWBGLEC-SYZQJQIISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mycophenolate mofetil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Mycophenolate mofetil is a prodrug of mycophenolic acid (Cat. # M5255) that is cleaved by nonspecific esterases in vivo to produce the parent compound. Mycophenolic acid blocks inosine monophosphate dehydrogenase and is a potent immunosuppresive agent.
Mycophenolate mofetil is a prodrug of mycophenolic acid and potent immunosuppresive agent.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eduardo F Mysler et al.
Arthritis and rheumatism, 65(9), 2368-2379 (2013-06-07)
To investigate the efficacy and safety of ocrelizumab in patients with class III/IV lupus nephritis (LN). Patients were randomized 1:1:1 to receive placebo, 400 mg ocrelizumab, or 1,000 mg ocrelizumab given as an intravenous infusion on days 1 and 15
K Budde et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 14(12), 2796-2806 (2014-10-04)
This Phase III randomized trial examined efficacy and safety of a novel once-daily extended-release tacrolimus formulation (LCP-Tacro [LCPT]) versus twice-daily tacrolimus in de novo kidney transplantation. Primary efficacy end point was proportion of patients with treatment failure (death, graft failure
John Seal et al.
Transplantation, 99(3), 623-628 (2014-08-26)
There is a paucity of contemporary data describing the results of pancreas retransplantation (PRT). As a measure of utility, we wished to determine whether PRT could produce equivalent short-term and long-term recipient outcomes to primary pancreas after kidney (PAK) transplantation.
Kremena V Star et al.
The American journal of surgical pathology, 37(9), 1319-1328 (2013-10-01)
Mycophenolate mofetil (MMF) is a T-cell inhibitor frequently used in the treatment of acute allograft rejection. MMF may cause colitis that clinically and histologically resembles graft-versus-host disease (GVHD). The aim of this study was to evaluate a wide range of
Christopher J Yates et al.
Transplantation, 96(8), 726-731 (2013-08-02)
The sensitivity of fasting plasma glucose (FPG) in screening for new-onset diabetes after transplantation (NODAT) has been questioned, particularly in the presence of moderate-dose prednisolone, where peak plasma glucose occurs 7 to 8 hr after administration. Oral glucose tolerance testing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service