Skip to Content
Merck
All Photos(2)

Key Documents

50744

Supelco

Dimethyl fumarate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH=CHCOOCH3
CAS Number:
Molecular Weight:
144.13
Beilstein:
774590
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

192-193 °C (lit.)

mp

102-106 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

[H]\C(=C(\[H])C(=O)OC)C(=O)OC

InChI

1S/C6H8O4/c1-9-5(7)3-4-6(8)10-2/h3-4H,1-2H3/b4-3+

InChI key

LDCRTTXIJACKKU-ONEGZZNKSA-N

Gene Information

human ... KEAP1(9817)

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl fumarate is a novel oral therapeutic agent, which can be used for patients suffering from relapsing-remitting multiple sclerosis.[1][2] It shows an effective inhibition of mould in bread[3] and can also serve as a potential candidate for the treatment of psoriasis, a chronic autoimmune condition.[2]
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effects of dimethyl fumarate on neuroprotection and immunomodulation
Albrecht P, et al.
Journal of Neuroinflammation, 163, 9-9 (2012)
Shoe contact dermatitis from dimethyl fumarate: clinical manifestations, patch test results, chemical analysis, and source of exposure
Gimenez-Arnau A, et al.
Contact Dermatitis, 61, 249-260 (2009)
Reactivity of dimethyl fumarate and methylhydrogen fumarate towards glutathione and N-acetyl-L-cysteine?preparation of S-substituted thiosuccinic acid esters
Schmidt.JT, et al.
Bioorganic & Medicinal Chemistry, 15, 333-342 (2007)
[Immunomodulation with oral dosage forms and their mechanisms of action].
R Gold
Drug research, 63 Suppl 1, S9-S9 (2013-11-01)
Sergey Kalinin et al.
Journal of neuroimmunology, 263(1-2), 13-19 (2013-08-07)
We previously showed that dimethyl fumarate (DMF) reduces inflammatory activation in astrocytes, involving activation of transcription factor Nrf2. However, the pathways causing Nrf2 activation were not examined. We now show that DMF modifies expression of histone deacetylases (HDACs) in primary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service