Skip to Content
Merck
All Photos(1)

Key Documents

Y0000336

Thiamazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methimazole, 1-Methyl-2-imidazolethiol, 2-Mercapto-1-methylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
Beilstein:
108646
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thiamazole

manufacturer/tradename

EDQM

mp

144-147 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1C=CNC1=S

InChI

1S/C4H6N2S/c1-6-3-2-5-4(6)7/h2-3H,1H3,(H,5,7)

InChI key

PMRYVIKBURPHAH-UHFFFAOYSA-N

Gene Information

human ... TPO(7173)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thiamazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Clementi et al.
American journal of medical genetics, 83(1), 43-46 (1999-03-17)
We report on a further case of congenital anomalies in a child exposed to methimazole during the first trimester of pregnancy (from first to seventh gestational week), and define a specific malformation pattern related to prenatal methimazole exposure and consisting
P Barbero et al.
American journal of medical genetics. Part A, 129A(1), 83-86 (2004-07-22)
We report three patients with bilateral choanal atresia in children prenatally exposed to methimazole (MMI) in order to define a MMI embryopathy clinical pattern. The combination of choanal atresia and other specific malformations strongly resembles previously reported patients exposed to
Fernando Bolaños et al.
Revista de investigacion clinica; organo del Hospital de Enfermedades de la Nutricion, 54(4), 307-310 (2002-11-06)
Long-term thionamide treatment is considered one of the main resources for Graves' hyperthyroidism. Although a 54.2% remission rate in patients so treated in Mexico was previously reported, most articles have shown a wide variation over time. In the present article
K E Hill et al.
New Zealand veterinary journal, 62(4), 208-213 (2013-12-20)
To determine the pharmacokinetics of a novel lipophilic formulation of transdermal methimazole compared to oral carbimazole. Healthy cats received 5 mg carbimazole orally every 12 hours for 13 treatments (n=6), then received transdermal methimazole (n=5) at a dose of 5 mg, then
Jeanne M Chattaway et al.
The Annals of pharmacotherapy, 41(6), 1018-1022 (2007-05-17)
To evaluate the evidence supporting the use of propylthiouracil (PTU) versus methimazole for the treatment of Graves' disease during pregnancy. An English-language literature search was conducted using MEDLINE (1966-March 2007). Identified articles were then reviewed for additional sources. Search terms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service