Skip to Content
Merck
All Photos(1)

Key Documents

SML0931

Sigma-Aldrich

Carbimazole

≥98% (HPLC)

Synonym(s):

1-Ethoxycarbonyl-3-methyl-2-thio-4-imidazoline, 1-Methyl-3-carbethoxy-2-thioglyoxalone, 2,3-Dihydro-3-methyl-2-thioxo-1H-imidazole-1-carboxylic acid ethyl ester, Athyromazole, Carbethoxymethimazole, Carbimazol, Ethyl 3-methyl-2-thioimidazoline-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O2S
CAS Number:
Molecular Weight:
186.23
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C7H10N2O2S/c1-3-11-7(10)9-5-4-8(2)6(9)12/h4-5H,3H2,1-2H3

InChI key

CFOYWRHIYXMDOT-UHFFFAOYSA-N

Biochem/physiol Actions

Carbimazole is a pro-drug of methimazole that is an inhibitor of thyroid enzyme thyroperoxidase. Carbimazole is used to treat hyperthyroidism. Also, carbimazole induces phosphorylation of eukaryotic elongation factor eEF2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodinated contrast-induced thyrotoxicosis.
Paul Dunne et al.
CMAJ : Canadian Medical Association journal = journal de l'Association medicale canadienne, 185(2), 144-147 (2012-11-14)
Franz Ludwig Dumoulin et al.
BMJ case reports, 2012, doi:10-doi:10 (2012-05-19)
A middle aged woman, previously healthy with the exception of mild seasonal asthma was presented with signs of acute hepatitis. The further investigation showed acute hepatitis E virus infection associated with autoimmune thyroiditis. Treatment was started with propranolol and carbimazol
E T Bell et al.
The Journal of small animal practice, 53(11), 661-663 (2012-09-11)
A 12-year-old female neutered ragdoll crossbred cat was presented for investigation of generalised weakness and regurgitation. The cat was being treated with transdermal methimazole for hyper-thyroidism, which had been diagnosed 10 weeks previously. An acetylcholine receptor antibody titre was consistent
Adoree Yi Ying Lim et al.
Singapore medical journal, 54(7), e133-e136 (2013-08-01)
A 24-year-old Chinese woman with Graves' disease presented with myositis two months after treatment with carbimazole. The patient's myositis resolved with hydration and cessation of carbimazole. No other causes of myositis were found, and a change in the medication to
[59 year-old man with skin and eye alterations and diabetes mellitus].
N Ewald et al.
Deutsche medizinische Wochenschrift (1946), 137(19), 997-998 (2012-05-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service