Skip to Content
Merck
All Photos(1)

Key Documents

Y0000333

Tiamulin hydrogen fumarate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tiamulin fumarate, Thiamutilin fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H51NO8S
CAS Number:
Molecular Weight:
609.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tiamulin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)[C@@]23CC[C@@H](C)[C@]1(C)[C@@H]2C(=O)CC3

InChI

1S/C28H47NO4S.C4H4O4/c1-8-26(6)17-22(33-23(31)18-34-16-15-29(9-2)10-3)27(7)19(4)11-13-28(20(5)25(26)32)14-12-21(30)24(27)28;5-3(6)1-2-4(7)8/h8,19-20,22,24-25,32H,1,9-18H2,2-7H3;1-2H,(H,5,6)(H,7,8)/b;2-1+/t19?,20-,22+,24-,25-,26+,27-,28-;/m0./s1

InChI key

YXQXDXAHCSEVSD-IHAONTCTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tiamulin hydrogen fumarate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K M S Islam et al.
Poultry science, 88(11), 2353-2359 (2009-10-17)
Tiamulin hydrogen fumarate is a semisynthetic derivative of the diterpene antibiotic pleuromutilin used in poultry medicine to treat mainly Mycoplasma- and Brachyspira-related diseases. Its use over 30 yr has not generally increased the development of resistance to these pathogens but
Petra Reinhold et al.
Veterinary journal (London, England : 1997), 187(3), 405-407 (2010-08-31)
The effect of short-term antimicrobial treatment on natural excretion of Chlamydia suis in rectal swabs and C. suis and Chlamydophila psittaci in nasal swabs was investigated in 47 clinically normal piglets by quantitative real-time PCR. Pigs were treated IM with
K M S Islam et al.
Poultry science, 87(12), 2528-2534 (2008-11-29)
In an earlier study, the continuous medication of broiler feed with a combination of tiamulin (TIA; 20 mg/kg), chlortetracycline (CTC; 60 mg/kg), and the ionophore anticoccidial salinomycin (SAL; 60 mg/kg) caused an initial increase in BW and feed efficiency (FE;
Xun Pan et al.
Journal of environmental sciences (China), 23(10), 1729-1737 (2011-01-01)
This work describes a systematic approach to the development of a method for simultaneous determination of three classes of veterinary antibiotics in the suspended solids (SS) of swine wastewater, including five sulfonamides, three tetracyclines and one macrolide (tiamulin). The entire
Saad Gharaibeh et al.
Veterinary microbiology, 150(3-4), 379-383 (2011-03-09)
This study compares the antimicrobial susceptibility over time between two groups of Mycoplasma gallisepticum (MG) isolates from the same geographical area. Minimum inhibitory concentration of 13 antimicrobials was determined against two groups of MG isolates from chickens. Group 1 strains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service