Skip to Content
Merck
All Photos(2)

Key Documents

34044

Supelco

Tiamulin

VETRANAL®, analytical standard

Synonym(s):

(1S,2R,3S,4S,6R,7S,14R)-6-[(2-Diethylaminoethylthio)acetoxy]-3-hydroxy-2,4,7,14-tetramethyl-4-vinyltricyclo[5,4,3,01.8]tetradecan-9-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H47NO4S
CAS Number:
Molecular Weight:
493.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)[C@@]23CC[C@@H](C)C1(C)[C@@H]2C(=O)CC3

InChI

1S/C28H47NO4S/c1-8-26(6)17-22(33-23(31)18-34-16-15-29(9-2)10-3)27(7)19(4)11-13-28(20(5)25(26)32)14-12-21(30)24(27)28/h8,19-20,22,24-25,32H,1,9-18H2,2-7H3/t19-,20+,22-,24+,25+,26-,27+,28+/m1/s1

InChI key

UURAUHCOJAIIRQ-QGLSALSOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide
Tiamulinis a veterinary drug with very low water solubility (0.000524 mg/mL). Its mode of action involves suppressing protein synthesis in bacteria.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
For detailed application information, please see the Frequently Asked Questions (FAQ) section at the bottom of this page.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Inhibition of peptide bond formation by pleuromutilins: the structure of the 50S ribosomal subunit from Deinococcus radiodurans in complex with tiamulin?
Schlunzen F, et al.
Molecular Microbiology, 54(05), 1287-1294 (2004)
Decreased susceptibility of MRSA ST398 to tiamulin.
Joseph E Rubin et al.
Veterinary microbiology, 151(3-4), 422-423 (2011-04-23)
Bénédicte F Callens et al.
Microbial drug resistance (Larchmont, N.Y.), 19(2), 146-151 (2012-12-20)
Streptococcus suis (S. suis) has often been reported as an important swine pathogen and is considered as a new emerging zoonotic agent. Consequently, it is important to be informed on its susceptibility to antimicrobial agents. In the current study, the
Güliz Gürel et al.
Journal of molecular biology, 389(1), 146-156 (2009-04-14)
Structures have been obtained for the complexes that tiamulin, homoharringtonine, and bruceantin form with the large ribosomal subunit of Haloarcula marismortui at resolutions ranging from 2.65 to 3.2 A. They show that all these inhibitors block protein synthesis by competing
Petra Reinhold et al.
Veterinary journal (London, England : 1997), 187(3), 405-407 (2010-08-31)
The effect of short-term antimicrobial treatment on natural excretion of Chlamydia suis in rectal swabs and C. suis and Chlamydophila psittaci in nasal swabs was investigated in 47 clinically normal piglets by quantitative real-time PCR. Pigs were treated IM with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service