Skip to Content
Merck
All Photos(1)

Documents

Y0000284

Fluticasone propionate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(6α,11β,16α,17α)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid S-(fluoromethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H31F3O5S
CAS Number:
Molecular Weight:
500.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluticasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF

InChI

1S/C25H31F3O5S/c1-5-20(31)33-25(21(32)34-12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h6-7,9,13,15-16,18-19,30H,5,8,10-12H2,1-4H3/t13-,15+,16+,18+,19+,22+,23+,24+,25+/m1/s1

InChI key

WMWTYOKRWGGJOA-CENSZEJFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluticasone propionate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Fluticasone propionate is a second generation glucocorticoid. Used as an anti-inflammatory agent for asthma. Shown to enhance eosinophil apoptosis in a concentration-dependent manner via the glucocorticoid receptor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Endocrine system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fouad J Moawad et al.
The American journal of gastroenterology, 108(3), 366-372 (2013-02-13)
Patients with clinical symptoms of esophageal dysfunction and dense eosinophilic infiltration of the esophageal mucosa are suspected to have eosinophilic esophagitis (EoE). Topical steroids are often used as first-line therapy for EoE, although some patients respond clinically to proton pump
Sanjeeva Dissanayake et al.
Respiratory medicine, 106 Suppl 1, S20-S28 (2013-01-01)
International asthma management guidelines recommend a long-acting β(2)-agonist (LABA) as add-on therapy in patients whose asthma is not controlled by low-dose inhaled corticosteroid (ICS) monotherapy. Treatment with a single inhaler containing an ICS/LABA combination is advocated because it may facilitate
Kate McKeage
Drugs, 73(2), 195-206 (2013-02-12)
The corticosteroid fluticasone propionate (fluticasone) and the long-acting β₂-adrenoceptor agonist formoterol fumarate (formoterol) have been combined in a single, pressurized, metered-dose, aerosol inhaler for the maintenance treatment of patients aged ≥12 years with persistent asthma. This article reviews the clinical
Melanie Manetsch et al.
PloS one, 8(3), e59635-e59635 (2013-03-28)
Mitogen-activated protein kinase phosphatase 1 (MKP-1) represses MAPK-driven signalling and plays an important anti-inflammatory role in asthma and airway remodelling. Although MKP-1 is corticosteroid-responsive and increased by cAMP-mediated signalling, the upregulation of this critical anti-inflammatory protein by long-acting β2-agonists and
David Price et al.
The Journal of allergy and clinical immunology, 131(3), 763-771 (2013-02-06)
Many asthmatic patients are unable to quit cigarettes; therefore information is needed on treatment options for smokers. This study evaluates 10 mg/d montelukast and 250 μg of fluticasone propionate twice daily, each compared with placebo, in patients with self-reported active smoking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service