Skip to Content
Merck
All Photos(1)

Key Documents

Y0000108

Trihexyphenidyl hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-α-Cyclohexyl-α-phenyl-1-piperidinepropanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31NO · HCl
CAS Number:
Molecular Weight:
337.93
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trihexyphenidyl hydrochloride

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.OC(CCN1CCCCC1)(C2CCCCC2)c3ccccc3

InChI

1S/C20H31NO.ClH/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21-15-8-3-9-16-21;/h1,4-5,10-11,19,22H,2-3,6-9,12-17H2;1H

InChI key

QDWJJTJNXAKQKD-UHFFFAOYSA-N

Gene Information

human ... CHRM1(1128)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trihexyphenidyl hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Case report: a case of anterocollis ameliorated by trihexyphenidyl].
Ayako Shioya et al.
Nihon Naika Gakkai zasshi. The Journal of the Japanese Society of Internal Medicine, 100(11), 3325-3328 (2012-01-19)
Parkinsonism complicating acute organophosphate insecticide poisoning.
Hasnur Zaman Hashim et al.
Annals of the Academy of Medicine, Singapore, 40(3), 150-151 (2011-05-24)
Bradley James Robottom et al.
The neurologist, 17(6), 340-341 (2011-11-03)
Trihexyphenidyl is 1 of the most effective agents for treatment of young-onset dystonia. As such, women of childbearing potential use trihexyphenidyl despite inadequate information about potential effects on pregnancy, labor, and fetal development. We report 2 uncomplicated pregnancies in 1
Yasumasa Yoshiyama et al.
Neurobiology of disease, 45(1), 329-336 (2011-09-06)
Anticholinergics, and drugs with anticholinergic properties, are widely and frequently prescribed, especially to the elderly. It is well known that these drugs decrease cognitive function and increase the risk of dementia. Although the mechanism of anticholinergic drug-induced cognitive impairment has
Alex Pollock et al.
The Cochrane database of systematic reviews, (10)(10), CD008389-CD008389 (2011-10-07)
Eye movement disorders may affect over 70% of stroke patients. These eye movement disorders can result in difficulty maintaining the normal ocular position and difficulty moving the eyes appropriately. The resulting functional disabilities include a loss of depth perception, reduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service