Skip to Content
Merck
All Photos(1)

Key Documents

Y0000094

Diosmin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3′,5,7-Trihydroxy-4′-methoxyflavone 7-rutinoside, Barosmin, Buchu resin, Diosmetin 7-O-rhamnosylglucoside, Diosmetin 7-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32O15
CAS Number:
Molecular Weight:
608.54
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

diosmin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2

InChI

1S/C28H32O15/c1-10-21(32)23(34)25(36)27(40-10)39-9-19-22(33)24(35)26(37)28(43-19)41-12-6-14(30)20-15(31)8-17(42-18(20)7-12)11-3-4-16(38-2)13(29)5-11/h3-8,10,19,21-30,32-37H,9H2,1-2H3/t10-,19+,21-,22+,23+,24-,25+,26+,27+,28+/m0/s1

InChI key

GZSOSUNBTXMUFQ-YFAPSIMESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diosmin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Citrus flavonoid. Modification of hesperidin. Studied for its chemopreventive properties, including anti-proliferative and anti-inflammatory.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Anna Bogucka-Kocka et al.
Natural product communications, 8(4), 545-550 (2013-06-07)
Diosmin is a naturally occurring flavone glycoside used in the treatment of venous diseases. In this review, we present the clinical aspects of the use of diosmin preparations in venous stasis, hemorrheologic disorders and vein wall remodeling. Because of its
Petr Vrbata et al.
International journal of pharmaceutics, 473(1-2), 407-413 (2014-07-30)
A nanofibrous membrane carrier for nearly water insoluble drug diosmin was formulated. The aim of this study was to evaluate the drug release and dissolution properties in an aqueous buffer of pH 7.8, and to compare the suitability of the
Monograph. Diosmin.
Alternative medicine review : a journal of clinical therapeutic, 9(3), 308-311 (2004-09-25)
Prasad Neerati et al.
Pharmacological reports : PR, 67(2), 339-344 (2015-02-26)
Diosmin is a natural flavone glycoside, a potent P-glycoprotein (P-gp) inhibitor in cultured cells and have the potential to alter the bioavailability of P-gp substrate drugs. However, the interaction between diosmin and fexofenadine is unreported. Hence, the present study was
Oomaidurai Senthamizhselvan et al.
European journal of pharmacology, 736, 131-137 (2014-04-29)
Reperfusion of ischemic tissue leads to the generation of oxygen derived free radicals which plays an important role in cellular damage. Objective of the current study is to evaluate the cardio-protective and antioxidant effect of diosmin on ischemia-reperfusion related cardiac

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service