Skip to Content
Merck
Diosmin analytical standard

Diosmin

Diosmin analytical standard
Synonym(s):
3′,5,7-Trihydroxy-4′-methoxyflavone 7-rutinoside, Barosmin, Buchu resin, Diosmetin 7-O-rhamnosylglucoside, Diosmetin 7-rutinoside, Diosmin
Empirical Formula (Hill Notation):
C28H32O15
CAS Number:
Molecular Weight:
608.54
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique