1.08411
L-Threonine
for biochemistry
Synonym(s):
L-Threonine, Thr, 2-Amino-3-hydroxybutanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H9NO3
CAS Number:
Molecular Weight:
119.12
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-774-1
NACRES:
NA.21
Recommended Products
Quality Level
form
solid
pH
5-6 (20 °C, 100 g/L in H2O)
mp
265-270 °C
solubility
90 g/L
bulk density
370 kg/m3
storage temp.
2-30°C
SMILES string
[N+H3]C(C(O)C)C(=O)[O-]
InChI
1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)
InChI key
AYFVYJQAPQTCCC-UHFFFAOYSA-N
Analysis Note
Assay (perchloric acid titration): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine powder
Appearance of solution (1 mol/l, water): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l): -15.4 to -16.4
Heavy metals (as Pb): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.01 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, fine powder
Appearance of solution (1 mol/l, water): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l): -15.4 to -16.4
Heavy metals (as Pb): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.01 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service