Skip to Content
Merck
All Photos(1)

Key Documents

T0500000

Tetracaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester, Amethocaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O2 · HCl
CAS Number:
Molecular Weight:
300.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tetracaine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CCCCNc1ccc(cc1)C(=O)OCCN(C)C

InChI

1S/C15H24N2O2.ClH/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3;/h6-9,16H,4-5,10-12H2,1-3H3;1H

InChI key

PPWHTZKZQNXVAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Topical ophthalmic anesthetic; used for spinal anesthesia

Biochem/physiol Actions

Blocks voltage-sensitive release of Ca2+ from sarcoplasmic reticulum.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akihiro Tanaka et al.
Antimicrobial agents and chemotherapy, 58(8), 4380-4383 (2014-05-21)
We have reported significantly higher convulsion prevalence in patients treated with cefepime than in those treated with meropenem. Additionally, cefepime-associated convulsions were found only in patients with brain disorders, not renal failure. Here, we compared the convulsive liability of cefepime
Halima Maulidi et al.
Pediatrics, 130(6), e1704-e1707 (2012-11-07)
Infants in NICUs undergo a variety of painful procedures. The management of pain has become an integral part of newborn infant care with the use of both systemic and topical agents to provide analgesia and anesthesia for procedural pain. Tetracaine
Yue Jiang et al.
International journal of pharmaceutics, 436(1-2), 806-814 (2012-08-09)
The aggregation behavior of the cationic drug/anionic surfactant vesicles formed by tetracaine hydrochloride (TH) and double-chain surfactant, sodium bis(2-ethylhexyl)sulfosuccinate (AOT), was investigated. By controlling the molar ratio of TH to AOT, a transition from catanionic vesicles to micelles was observed.
Thomas B Marriott et al.
Clinical therapeutics, 34(10), 2174-2183 (2012-09-05)
The lidocaine/tetracaine heated patch is typically applied to the skin for 20 to 30 minutes to provide local dermal analgesia prior to venous access or minor dermatologic procedures. The potential exists for the use of multiple heated patches for longer
Quan Wen et al.
Cornea, 32(2), 179-184 (2012-08-08)
Our recent tissue cross-linking studies have raised the possibility of using aliphatic β-nitroalcohols (BNAs) for pharmacologic, therapeutic corneal cross-linking. The present study was performed to determine the permeability of BNAs and to explore the use of permeability-enhancing agents. Ex vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service