Skip to Content
Merck
All Photos(1)

Key Documents

S5632

Supelco

Sulfamethizole

analytical standard, ≥99% (HPLC)

Synonym(s):

4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzenesulfonamide, N1-(5-Methyl-1,3,4-thiadiazol-2-yl)sulfanilamide, Sulfamethylthiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N4O2S2
CAS Number:
Molecular Weight:
270.33
Beilstein:
255002
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

Assay

≥99% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

Cc1nnc(NS(=O)(=O)c2ccc(N)cc2)s1

InChI

1S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5H,10H2,1H3,(H,12,13)

InChI key

VACCAVUAMIDAGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfamethizole is an antibacterial drug used in the treatment of the urinary tract infections.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfamethizole may be used as a reference standard to examine the antibiotic remnants in honey by multianalyte/multiclass ultra-performance liquid chromatography–tandem mass spectrometry (UPLC–MS/MS) method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coordination chemistry of sulfamethizole: crystal structures of [Cu (sulfamethizolate) 2 (py) 2 (OH 2)]? H 2 O,[M (sulfamethizolate) 2 (py) 2 (OH 2) 2][M= Co and Ni] and {Cu (sulfamethizolate) 2 (dmf) 2}?.
Borras E, et al.
Polyhedron, 19(15), 1859-1866 (2000)
Development of a rapid method for the determination of antibiotic residues in honey using UPLC-ESI-MS/MS.
Kivrak I, et al.
Food Sci. Technol., 36(1), 90-96 (2016)
Tony L Yaksh et al.
Anesthesiology, 108(5), 938-949 (2008-04-24)
Intrathecal N-methyl-d-aspartate antagonists have antihyperalgesic efficacy. The authors examined toxicity in a canine model of chronic lumbar intrathecal infusion. Dogs (10-16 kg) were prepared with lumbar intrathecal catheters connected to vest-mounted pumps (100 microl/h). In phase 1, stepwise incrementations in
Ali Md Ahad et al.
Bioorganic & medicinal chemistry, 19(6), 2046-2054 (2011-03-01)
Disruption of the phosphatidylinositol 3-kinase/AKT signaling pathway can lead to apoptosis in cancer cells. Previously we identified a lead sulfonamide that selectively bound to the pleckstrin homology (PH) domain of AKT and induced apoptosis when present at low micromolar concentrations.
Robert R Muder
Seminars in respiratory and critical care medicine, 28(6), 672-677 (2007-12-21)
Stenotrophomonas (Xanthomonas) maltophilia is a nonfermentative, gram-negative bacillus that is widely distributed in natural and human-made environments. In the nonhospital setting it is an uncommon pathogen, typically causing soft tissue infection of contaminated wounds. In the hospital setting, particularly among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service