Skip to Content
Merck
All Photos(3)

Key Documents

S6422

Sigma-Aldrich

Sodium pyrophosphate tetrabasic decahydrate

BioXtra, 99.0-103.0%

Synonym(s):

Pyrophosphate inorganic, Sodium diphosphate tetrabasic, Tetrasodium pyrophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na4P2O7 · 10H2O
CAS Number:
Molecular Weight:
446.06
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

99.0-103.0%

form

powder

impurities

≤0.01% Insoluble matter

pH

9.5-10.5 (20 °C, 50 g/L)

solubility

H2O: 0.5 M, clear, colorless

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O

InChI

1S/4Na.H4O7P2.10H2O/c;;;;1-8(2,3)7-9(4,5)6;;;;;;;;;;/h;;;;(H2,1,2,3)(H2,4,5,6);10*1H2/q4*+1;;;;;;;;;;;/p-4

InChI key

VZWGHDYJGOMEKT-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Sodium pyrophosphate tetrabasic decahydrate functions as a sequestering, clarifying, buffering agent and water softener. It is used as an emulsifier, pH buffer and dough conditioner. Sodium pyrophosphate tetrabasic acts as a thickening agent in puddings and tartar control agent in toothpaste.

Application

Sodium pyrophosphate tetrabasic decahydrate has been used:
  • for pyrophosphate hydrolysis and quantification
  • as a component of extraction buffer to homogenize liver
  • as a component of cell lysis buffer for western blotting

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Why There's Antifreeze in Your Toothpaste: The Chemistry of Household Ingredients (2007)
Defective lysosomal clearance of autophagosomes and its clinical implications in nonalcoholic steatohepatitis
Wang X, et al.
Faseb Journal, 32(1), 37-51 (2017)
A Consumerys Dictionary of Household, Yard and Office Chemicals: Complete Information About Harmful and Desirable Chemicals Found in Everyday Home Products, Yard Poisons, and Office Polluters (2007)
Leptin, BMI, and a metabolic gene expression signature associated with clinical outcome to VEGF inhibition in colorectal cancer
Pommier AJC, et al.
Cell Metabolism, 23(1), 77-93 (2016)
Hydrolysis of Extracellular Pyrophosphate increases in post-hemodialysis plasma
Azpiazu D, et al.
Scientific reports, 8 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service