Skip to Content
Merck
All Photos(1)

Key Documents

S0900000

Sodium taurocholate

BRP, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Taurocholic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44NNaO7S
CAS Number:
Molecular Weight:
537.68
Beilstein:
3901620
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sodium taurocholate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[NaH].[S](=O)(=O)(O)CCNC(=O)CC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@H](C[C@H]3O)C[C@@H](CC4)O)C)C[C@@H]2O)CC1)C)C

InChI

1S/C26H45NO7S.Na.H/c1-15(4-7-23(31)27-10-11-35(32,33)34)18-5-6-19-24-20(14-22(30)26(18,19)3)25(2)9-8-17(28)12-16(25)13-21(24)29;;/h15-22,24,28-30H,4-14H2,1-3H3,(H,27,31)(H,32,33,34);;/t15-,16+,17-,18-,19+,20+,21-,22+,24+,25+,26-;;/m1../s1

InChI key

VXRNGJZKPMKERN-NEMAEHQESA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Anionic detergent used for protein solubilization.

Biochem/physiol Actions

Non-sulfated bile salt, physiological transport substrate for the bile salt export pump/sister of Pgp (BSEP/spgp), Na(+)/taurocholate cotransporter (NTCP) and MRP3 into the hepatic carnalicular.
Non-sulfated bile salt.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp Uhl et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 108, 111-118 (2017-07-19)
Despite the outstanding progress in modern medicine, the oral delivery of peptide drugs is limited until today due to their instability in the gastrointestinal tract and low mucosa penetration. To overcome these hurdles, liposomes containing the specific tetraether lipid GCTE
Takashi Yoshikado et al.
Molecular pharmacology, 83(2), 552-562 (2012-12-12)
The antiplatelet drug, ticlopidine (TIC), reportedly causes cholestatic liver injuries. The present study analyzed the effect of TIC on bile formation, revealing that the biliary secretion of phospholipids was significantly decreased in TIC-administered Sprague Dawley (SD) rats. However, the effect
Antonella Zacheo et al.
Scientific reports, 10(1), 9052-9052 (2020-06-05)
Dengue virus (DENV) causes 390 million infections per year. Infections can be asymptomatic or range from mild fever to severe haemorrhagic fever and shock syndrome. Currently, no effective antivirals or safe universal vaccine is available. In the present work we
Hong-Yu Xu et al.
Colloids and surfaces. B, Biointerfaces, 181, 927-934 (2019-08-07)
Berberine (BBR) is an important natural product with poor gastrointestinal behavior includes low permeability, P-glycoprotein efflux, and mass elimination in the intestine. The aim of this study was to develop a novel nanoemulsion (NE) to improve the hypoglycemic efficacy of
Inha Heo et al.
Nature microbiology, 3(7), 814-823 (2018-06-28)
Stem-cell-derived organoids recapitulate in vivo physiology of their original tissues, representing valuable systems to model medical disorders such as infectious diseases. Cryptosporidium, a protozoan parasite, is a leading cause of diarrhoea and a major cause of child mortality worldwide. Drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service