Skip to Content
Merck
All Photos(1)

Key Documents

S0600000

Silicone oil

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-Si(CH3)2O-]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

>1 (vs air)

vapor pressure

<5 mmHg ( 25 °C)
5 mmHg ( 20 °C)

API family

silicone oil

manufacturer/tradename

EDQM

refractive index

n20/D 1.403 (lit.)

bp

>140 °C/0.002 mmHg (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Silicone oil EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael L Samsom et al.
Experimental eye research, 127, 14-19 (2014-07-06)
Proteoglycan 4 (PRG4, or lubricin) is a lubricating mucin-like glycoprotein recently discovered at the ocular surface, where it functions as a boundary lubricant and appears to play a protective role. Recent technological advances have enabled abundant expression of full-length recombinant
Kristan S Worthington et al.
Experimental eye research, 121, 194-199 (2014-03-13)
Sub-retinal implantation of foreign materials is becoming an increasingly common feature of novel therapies for retinal dysfunction. The ultimate compatibility of implants depends not only on their in vitro chemical compatibility, but also on how well the mechanical properties of the
Jessica H Wen et al.
Nature materials, 13(10), 979-987 (2014-08-12)
Stem cells regulate their fate by binding to, and contracting against, the extracellular matrix. Recently, it has been proposed that in addition to matrix stiffness and ligand type, the degree of coupling of fibrous protein to the surface of the
M C Prieto-Blanco et al.
The Science of the total environment, 503-504, 105-112 (2014-08-13)
This report describes a polydimethylsiloxane (PDMS)-thymol/nitroprusside delivery composite sensor for direct monitoring of ammonium in environmental water samples. The sensor is based on a PDMS support that contains the Berthelot's reaction reagents. To prepare the PDMS-thymol/nitroprusside composite discs, thymol and
Thomas-Benjamin Seiler et al.
Chemosphere, 112, 77-84 (2014-07-23)
As part of the risk assessment process within REACh, prior to manufacturing and distribution of chemical substances their (eco)toxicological impacts have to be investigated. The fish embryo toxicity test (FET) with the zebrafish Danio rerio has gained a high significance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service