Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicone oil
Linear Formula:
[-Si(CH
3
)
2
O-]
n
CAS Number:
63148-62-9
Products
Brand
Sigma-Aldrich (13)
Boiling Point (°C)
Physical Form
Quality Segment
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
317667
Silicone oil,
viscosity 5 cSt (25 °C)
Expand
View Pricing
378321
Silicone oil,
viscosity 10 cSt (25 °C)
Expand
View Pricing
378364
Silicone oil,
viscosity 100 cSt (25 °C)
Expand
View Pricing
378348
Silicone oil,
viscosity 20 cSt (25 °C)
Expand
View Pricing
378399
Silicone oil,
viscosity 1,000 cSt (25 °C)
Expand
View Pricing
85409
Silicone oil,
for oil baths (from −50°C to +200°C)
Expand
View Pricing
378372
Silicone oil,
viscosity 350 cSt (25 °C)
Expand
View Pricing
378380
Silicone oil,
viscosity 500 cSt (25 °C)
Expand
View Pricing
378356
Silicone oil,
viscosity 50 cSt (25 °C)
Expand
View Pricing
378402
Silicone oil,
viscosity 10,000 cSt (25 °C)
Expand
View Pricing
181838
Silicone oil,
viscosity 60,000 cSt (25 °C)
Expand
View Pricing
146153
Silicone oil,
for melting point and boiling point apparatuses
Expand
View Pricing
378437
Silicone oil,
viscosity 100,000 cSt (25 °C)
Expand
View Pricing
378429
Silicone oil,
viscosity 30,000 cSt (25 °C)
Expand
View Pricing
S0600000
Silicone oil,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing