Skip to Content
Merck
All Photos(2)

Key Documents

RDD039

Sigma-Aldrich

Imidazole

for molecular biology, ≥99% (titration), free-flowing, Redi-Dri

Synonym(s):

1, 3-diazole, 1,3-Diaza-2,4-cyclopentadiene, Glyoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103853
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

grade

for molecular biology

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

product line

Redi-Dri

Assay

≥99% (titration)

form

crystalline powder

quality

free-flowing

pH

6.2-7.8 (25 °C)

pKa (25 °C)

6.95

bp

256 °C (lit.)

mp

88-91 °C (lit.)

application(s)

diagnostic assay manufacturing

foreign activity

DNase, RNase and protease, none detected

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imidazole is a heterocyclic compound with a five-membered planar ring. It is amphoteric and highly polar.

Application

Excellent for buffers in the range of pH 6.2-7.8
Imidazole has been used
  • in the lysis, wash, and elution buffer for the purification of histidine tagged proteins
  • in gradient for the purification of histidine tagged proteins using nickel affinity chromatography
  • as a component of homogenization buffer for the purification of phagosomal compartments from dendritic cells

Biochem/physiol Actions

Imidazole derivatives have antibacterial, antifungal and anticancer functionality. It interacts with DNA and also binds to protein and stops cell division. It also acts as a microtubule destabilizing agents and inhibits topoisomerase and Cytochrome P450 Family 26 Subfamily A Member 1 (CYP26A1) enzymes. Imidazole based anticancer drug find applications in cancer chemotherapy. It is used as buffer component for purification of the histidine tagged recombinant proteins in immobilized metal-affinity chromatography (IMAC).

Features and Benefits

  • Qualified for molecular biology applications, DNase, Rnase, and protease free
  • Redi-Dri packaging controls moisture, eliminating clumps
  • Crystalline powder remains free-flowing and much easier to use
  • Safer to handle and weigh out for routine use in lab
  • Less waste, loss of drying, while maintaining high quality

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular pathways regulating pro-migratory effects of Hedgehog signaling
Hochman E, et al.
The Journal of Biological Chemistry, 281(45), 33860-33870 (2006)
Imidazoles as potential anticancer agents
Ali I, et al.
MedChemComm, 8(9), 1742-1773 (2017)
Functional expression of novel human and murine AKR1B genes
Salabei JK, et al.
Chemico-Biological Interactions, 191(1-3), 177-184 (2011)
Imidazole: Having versatile biological activities
Verma A, et al.
Journal of Chemistry, 2013 (2013)
[Purification of proteins using polyhistidine affinity tags
Bornhorst JA and Falke JJ
Methods in Enzymology, 326, 245-254 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service