Skip to Content
Merck
All Photos(2)

Documents

RDD028

Sigma-Aldrich

Ethanolamine hydrochloride

anhydrous, free-flowing, Redi-Dri, ≥99.0%

Synonym(s):

Colamine hydrochloride, Ethanolamine chloride, 2-Aminoethanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH2OH · HCl
CAS Number:
Molecular Weight:
97.54
Beilstein:
3542892
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

grade

anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99.0%

quality

free-flowing

pKa (25 °C)

9.5

mp

82-84 °C (lit.)

SMILES string

NCCO.[H]Cl

InChI

1S/C2H7NO.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

PMUNIMVZCACZBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethanolamine hydrochloride, a versatile compound in biochemistry and molecular biology, plays a pivotal role in diverse research areas. Primarily, it functions as a buffering agent, ensuring a stable pH environment crucial for enzymatic reactions, protein solubilization, and other biological processes. Its significance extends to membrane protein studies, aiding in the extraction of these proteins from cell membranes. It also finds utility in cell culture and as a reagent for biological sample preparation, simplifying nucleic acid and protein isolation, as well as facilitating the detection of proteins by western blotting and immunohistochemistry. This compound′s wide-ranging applications make it indispensable in biochemical research, contributing to various experimental procedures and advancing our understanding in fields such as molecular biology, immunology, and cellular biology.

Application

Redi-Dri is an innovative packaging product line that helps eliminate clumping material by adding extra packaging process steps without any chemical additives, anti-clumping agents, or hydrophobic compounds. Redi-Dri offers a more convenient and safer way to handle the products.
Ethanolamine hydrochloride has been used:
  • in overnight incubation of the tips to attach primary amine groups at the tip surface
  • in the preparation of DMEM (dulbecco′s modified eagle′s medium)/F-12 media to culture human epidermal growth factor receptor 2 (HER2) cells derived from MMTV-HER2 transgenic mouse mammary tumors
  • to administer the cultures to study its effect on the endogenous phosphatidyl ethanolamine pool and autophagy process

Biochem/physiol Actions

Ethanolamine hydrochloride is utilized as a carbon and nitrogen source by bacteria that differs phylogenetically. It exists as phosphotidylethanolamine in the bacterial and mammalian cell membrane. Ethanolamine-ammonia lyase is responsible for the degradation of ethanolamine into acetaldehyde and ammonia. Ethanolamine is known to positively support lipid accumulation in photosynthetic organism model.

Features and Benefits

  • Suitable for molecular biology
  • Redi-Dri packaging controls moisture for hygroscopic chemicals that commonly clump
  • Powder remains free-flowing and easy to use
  • Safer to handle and weigh out for routine use in the lab
  • Eliminates clumps, maintaining product moisture, quality, and product specifications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unbinding molecular recognition force maps of localized single receptor molecules by atomic force microscopy
Sotres J, et al.
ChemPhysChem, 9(4), 590-599 (2008)
K C Andree et al.
Lab on a chip, 19(6), 1006-1012 (2019-02-15)
The load of circulating tumor cells (CTC) is related to poor outcomes in cancer patients. A sufficient number of these cells would enable a full characterization of the cancer. An approach to probe larger blood volumes, allowing for the detection
Foodborne Diseases: New Insights for the Healthcare Professional, 14-14 (2012)
Selective cyclin-dependent kinase 2/cyclin A antagonists that differ from ATP site inhibitors block tumor growth
Mendoza N, et al.
Cancer Research, 63(5), 1020-1024 (2003)
Amino Alcohols-Advances in Research and Application, 29-29 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service