Skip to Content
Merck
All Photos(1)

Documents

15014

Sigma-Aldrich

Ethanolamine

≥99%

Synonym(s):

2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
Beilstein:
505944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.1 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

1436 °F

expl. lim.

17 %

impurities

≤0.5% water (Karl Fischer)

ign. residue

≤0.05% (as SO4)

refractive index

n20/D 1.454 (lit.)

bp

170 °C (lit.)
69-70 °C/10 mmHg

mp

10-11 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

cation traces

Fe: ≤10 mg/kg

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Catalytic performance and acidic analysis of chloroaluminate ionic liquid: Explores the effects of ethanolamine on the catalytic properties of ionic liquids in oil synthesis (Hu et al., 2024).

Other Notes

The article number 15014-4X2.5L will be discontinued. Please order the single bottle 15014-2.5L which is physically identical with the same exact specifications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Vance
Biochimica et biophysica acta, 1045(2), 128-134 (1990-07-16)
Monolayer cultures of rat hepatocytes have been examined for their ability to secrete ethanolamine plasmalogen as a component of nascent lipoproteins. In culture medium from these cells, ethanolamine plasmalogen comprises approx. 20-30% of total ethanolamine glycerophospholipids when measured either as
Gabriel da Silva
The journal of physical chemistry. A, 116(45), 10980-10986 (2012-09-22)
The alkanolamine 2-aminoethanol (NH(2)CH(2)CH(2)OH), otherwise known as monoethanolamine (MEA), is a widely used solvent for carbon capture, yet relatively little is known about its atmospheric chemistry. The hydroxyl radical initiated oxidation of MEA is thought to predominantly form the α-aminoalkyl
Ye-Jin Hwang et al.
Advanced materials (Deerfield Beach, Fla.), 27(31), 4578-4584 (2015-07-03)
By controlling the polymer/polymer blend self-organization rate, all-polymer solar cells composed of a high-mobility, crystalline, naphthalene diimide-selenophene copolymer acceptor and a benzodithiophene-thieno[3,4-b]thiophene copolymer donor are achieved with a record 7.7% power conversion efficiency and a record short-circuit current density (18.8
H Perschak et al.
Human neurobiology, 6(3), 191-194 (1987-01-01)
Concentrations of putative neuroactive substances glutamate, aspartate, gamma-aminobutyric acid, glycine, proline and ethanolamine were determined in ventricular cerebrospinal fluid collected in patients suffering from Parkinson's disease, pain syndromes or cerebellar tremor. Values are similar to those given in the literature
Su Chul Jang et al.
Small (Weinheim an der Bergstrasse, Germany), 11(4), 456-461 (2014-09-10)
Evaluation of kinetic distribution and behaviors of nanoparticles in vivo provides crucial clues into their roles in living organisms. Extracellular vesicles are evolutionary conserved nanoparticles, known to play important biological functions in intercellular, inter-species, and inter-kingdom communication. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service