Skip to Content
Merck
All Photos(1)

Key Documents

PHR1423

Supelco

Sodium Nitroprusside

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Sodium nitroferricyanide(III) dihydrate, Nitroprusside sodium, SNP, Sodium nitroprusside, Sodium pentacyanonitrosylferrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
297.95
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1614501

API family

sodium nitroprusside

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O=N[Fe--](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q;;;;;2*-1;2*+1;;

InChI key

OIRZWVYIQXBRFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Sodium Nitroprusside is widely used as a drug to manage hypertension, via lowering elevated blood pressure by acting on the arteriolar smooth muscle.

Application

Sodium Nitroprusside may be used as a spectrophotometric reagent for the analysis of meloxicam in pharmaceutical formulations using spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7818 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antihypertensive Drugs (1997)
Oliver Baretella et al.
Life sciences, 118(2), 206-212 (2014-01-15)
Levels of the endothelium-derived peptide endothelin-1 (ET-1) are elevated in obese humans, and ET-1 mediated vascular tone is increased. Renal arterial smooth muscle is highly responsive to ET-1. Whether or not endothelium-derived ET-1 affects contractions of the renal artery under
Rohit Ramchandra et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 307(3), R271-R280 (2014-05-23)
Heart failure (HF) is associated with increased cardiac and renal sympathetic drive, which are both independent predictors of poor prognosis. A candidate mechanism for the centrally mediated sympathoexcitation in HF is reduced synthesis of the inhibitory neuromodulator nitric oxide (NO)
Gary J Hodges et al.
Experimental physiology, 99(2), 381-392 (2013-11-12)
What is the central question of this study? Previous work has produced the counterintuitive finding that the vasoconstrictor neurotransmitters noradrenaline and neuropeptide Y are involved in vasodilatation. We aimed to discover whether sympathetic neurotransmitters are required for the sustained vasodilatation
Laura A E Cox et al.
Shock (Augusta, Ga.), 43(4), 322-326 (2015-01-08)
Although endothelial dysfunction is central to the pathogenesis of sepsis, no specific and clinically applicable marker for endothelial dysfunction is currently available. Endocan, a proteoglycan excreted by endothelial cells in response to inflammatory cytokines, may serve as such a marker.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service