Skip to Content
Merck
All Photos(1)

Key Documents

PHR1232

Supelco

Calcium-d-pantothenate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

D-Pantothenic acid hemicalcium salt, (R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27
Beilstein:
3769272
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C0400000
traceable to USP 1087009

API family

calcium pantothenate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)O[Ca]OC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1

InChI key

FAPWYRCQGJNNSJ-UBKPKTQASA-L

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Calcium-d-pantothenate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by ion chromatography.
Precursor in the biosynthesis of coenzyme A.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Physical properties

Due to the unstable, hygroscopic nature of the free acid, the calcium salt is employed.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3618 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Yoshimoto et al.
International journal of molecular sciences, 21(2) (2020-01-19)
To investigate environmental factors that contribute to ultraviolet A (UVA)-induced oxidative stress, which accelerates the senescence and toxicity of skin cells, we irradiated human fibroblasts cultured in commonly used essential media with UVA and evaluated their viability and production of
Kristina Riegel et al.
Clinical, cosmetic and investigational dermatology, 13, 499-510 (2020-08-18)
Clinical studies have confirmed that the hair-growth-promoting effect of approved oral drug combinations is beneficial for the treatment of diffuse telogen effluvium, which is characterized by the excessive loss of telogen club hairs. Since data elucidating the mode of action
Dietary supplements containing vitamins and minerals: Formulation, optimization and evaluation
Rajakumari R, et al.
Powder Technology, 336, 481-492 (2018)
My Tuyen Thi Nguyen et al.
Nutrients, 12(6) (2020-06-21)
Vitamins are the essential elements for human life and, particularly, for infant health. Human milk is the best source of nutrients for newborns, however, the information of vitamins in Asian maternal milk is still limited. In this study, we have
Chunwu Zhu et al.
Science advances, 4(5), eaaq1012-eaaq1012 (2018-05-29)
Declines of protein and minerals essential for humans, including iron and zinc, have been reported for crops in response to rising atmospheric carbon dioxide concentration, [CO2]. For the current century, estimates of the potential human health impact of these declines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service