Skip to Content
Merck
All Photos(1)

Key Documents

PHL89270

Rutin

phyproof® Reference Substance

Synonym(s):

3,3′,4′,5,7-Pentahydroxyflavone 3-rutinoside, 3-Rutinosylquercetin, Globulariacitrin, Ilixanthi, Myrticalorin, Osyritrin, Paliuroside, Phytomelin, Quercetin 3-rhamnoglucoside, Quercetin 3-rutinoside, Rutoside, Sophorin, Tanrutin, Violaquercitrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O16
CAS Number:
Molecular Weight:
610.52
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

O1[C@H]([C@@H]([C@@H]([C@H]([C@@H]1C)O)O)O)OC[C@H]2O[C@H]([C@@H]([C@H]([C@@H]2O)O)O)Oc3[c](c4c([o]c3c5cc(c(cc5)O)O)cc(cc4O)O)=O

InChI

1S/C27H30O16/c1-8-17(32)20(35)22(37)26(40-8)39-7-15-18(33)21(36)23(38)27(42-15)43-25-19(34)16-13(31)5-10(28)6-14(16)41-24(25)9-2-3-11(29)12(30)4-9/h2-6,8,15,17-18,20-23,26-33,35-38H,7H2,1H3/t8-,15+,17-,18+,20+,21-,22+,23+,26+,27-/m0/s1

InChI key

IKGXIBQEEMLURG-NVPNHPEKSA-N

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, and inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG.

Rutin, also known as vitamin P, is an abundantly found polyphenolic flavonoid, known for its anti-inflammatory, antitumor, and anticancer properties.

Application

This primary reference standard can also be used as follows:
  • Electrochemical determination of rutin using a glassy carbon electrode (GCE), modified by a copper-complexed chitosan/multiwalled carbon nanotubes (Cu-CS/MWCNT) in fruit samples of orange, lemon, lime, and apple
  • Cyclic voltametric detection and quantification of rutin in pharmaceutical formulations using novel nickel nanoparticles-graphene oxide composite-glassy carbon electrode (Ni-GO/GCE)
  • Spectrofluorimetric quantification of rutin in a pharmaceutical sample using carbon dots (CDs) prepared from a non-essential amino acid precursor, L-asparagine
  • Analysis of human serum and pharmaceutical tablet samples for the estimation of rutin by using a porous carbon-cobalt tungstate (PC/CoWO4) nanosheet based electrochemical sensor
  • Simultaneous determination of rutin and isoquercitrin by differential pulse voltammetry (DPV) using glassy carbon electrode (GCE) modified with deposition of thio-β-cyclodextrin functionalized graphene/palladium nanoparticles (SH-β-CD-Gr/PdNPs) for pharmaceutical and medical analysis
  • Ionic liquid vacuum microwave-assisted extraction (ILVMAE) of rutin, hyperoside, and hesperidin from Sorbus tianschanica leaf samples for their subsequent quantification by HPLC
  • Simultaneous detection and quantification of vitexin, rutin, hyperoside, and quercetin in hawthorn flower and leaf extract samples using high-performance liquid chromatography combined with spectrophotometric detection (HPLC-UV)

Other Notes

This compound is commonly found in plants of the genus: achillea crataegus echinacea eucalyptus ginkgo glycyrrhiza hedera humulus hypericum lycium pimpinella primula sambucus serenoa

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abolfazl Alirezalu et al.
Foods (Basel, Switzerland), 9(4) (2020-04-09)
Hawthorn belongs to the Crataegus genus of the Rosaceae family and is an important medicinal plant. Due to its beneficial effects on the cardiovascular system and its antioxidant and antimicrobial activity hawthorn has recently become quite a popular herbal medicine
Zipora Tietel et al.
Scientific reports, 10(1), 14339-14339 (2020-09-02)
Worldwide demand for tef (Eragrostis tef) as a functional food for human consumption is increasing, thanks to its nutritional benefits and gluten-free properties. As a result, tef in now grown outside its native environment in Ethiopia and thus information is
Tshudufhadzo Mphaphuli et al.
Foods (Basel, Switzerland), 9(4) (2020-04-09)
Natal plum fruit (Carissa macrocarpa) is indigenous to South Africa and a rich source of cyanidin derivatives. Indigenous fruits play a major role in food diversification and sustaining food security in the Southern African region. Agro-processing of indigenous are practiced
Achillia Lakka et al.
Antioxidants (Basel, Switzerland), 8(12) (2019-11-30)
The current investigation was undertaken to examine saffron processing waste (SPW) as a bioresource, which could be valorized to produce extracts rich in antioxidant polyphenols, using a green, natural deep eutectic solvent (DES). Initially, there was an appraisal of the
Stefan Ivanović et al.
Foods (Basel, Switzerland), 9(4) (2020-04-09)
The present study evaluates natural composition of Serbian roasted hazelnut skins (HS) with potential role in application as functional nutrient of various food products. Total phenols (TPC) and flavonoids contents (TFC) in HS extracts obtained with different ethanol concentrations (10%-I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service