Skip to Content
Merck
All Photos(1)

Documents

00740580

Quercitrin

primary reference standard

Synonym(s):

3,3′,4′,5,7-Pentahydroxyflavone 3-rhamnoside, 3-O-Rhamnosylquercetin, Quercetin 3-rhamnoside, Quercimelin, Quercitroside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
Beilstein:
68135
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

SMILES string

C[C@@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C21H20O11/c1-7-15(26)17(28)18(29)21(30-7)32-20-16(27)14-12(25)5-9(22)6-13(14)31-19(20)8-2-3-10(23)11(24)4-8/h2-7,15,17-18,21-26,28-29H,1H3/t7-,15-,17+,18+,21-/m0/s1

InChI key

OXGUCUVFOIWWQJ-HQBVPOQASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Other Notes

This compound is commonly found in plants of the genus: eucalyptus ginkgo hypericum mentha

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco Assis Bezerra da Cunha et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 614-621 (2016-10-04)
Eugenia uniflora is used in the Brazilian folk medicine to treat intestinal disorders and hypertension. However, scanty information exist on its potential toxicity to human, and little is known on its antioxidant activity in biological system. Hence, we investigated for
Hyeong-Geug Kim et al.
Frontiers in pharmacology, 11, 447-447 (2020-04-30)
Stress is a well-known factor for inflammation in diverse organs/tissues. Stress also leads to liver injury, which was supported by clinical observations and animal studies. We herein investigated the hepatoprotective property of an herbal formula (called as CGplus) consisting of
Xiaolan Jiang et al.
PloS one, 8(4), e62315-e62315 (2013-05-07)
Phenolic compounds in tea plant [Camellia sinensis (L.)] play a crucial role in dominating tea flavor and possess a number of key pharmacological benefits on human health. The present research aimed to study the profile of tissue-specific, development-dependent accumulation pattern
E A Cruz et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(2), 115-121 (2011-08-02)
Aqueous extract of Kalanchoe pinnata (Kp) have been found effective in models to reduce acute anaphylactic reactions. In the present study, we investigate the effect of Kp and the flavonoid quercetin (QE) and quercitrin (QI) on mast cell activation in
Farid Khallouki et al.
Food chemistry, 221, 1034-1040 (2016-12-17)
Previous studies have shown that Argan fruits contain a large variety of polyphenolic compounds. Recently, another class of polyphenolic compounds, namely amino phenols have been detected and identified in immature Argan fruits. The objective of this study, was to establish

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service