Skip to Content
Merck
All Photos(3)

Key Documents

P9458

Sigma-Aldrich

Potassium sulfate

BioXtra, ≥99.0%

Synonym(s):

K2SO4, Sulphate of potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
Assay:
≥99.0%
form:
powder
solubility:
H2O: 1 M, clear, colorless

product line

BioXtra

Quality Level

Assay

≥99.0%

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.001%

cation traces

Al: ≤0.0005%
Ca: ≤0.01%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.01%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium sulphate has been used as a component of fermentation medium for culturing Pichia pastoris, minimal medium for growing microbial cultures and magnesium transporter A (MgtA) purification buffers.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Absolute quantitation of intracellular metabolite concentrations by an isotope ratio-based approach
Bennett BD, et al.
Nature Protocols, 3(8), 1299-1299 (2008)
Heterologous expression and purification of the magnesium transporter A (MgtA) in Escherichia coli
Subramani S and Morth JP
Bio-protocol, 6(22), 1299-1299 (2016)
Production of recombinant protein in Pichia pastoris by fermentation
Tolner B, et al.
Nature Protocols, 1(2), 1006-1006 (2006)
Jinyong Yan et al.
Bioresource technology, 102(7), 4755-4758 (2011-02-01)
A dual modification procedure composed of cross-linking and protein coating with K(2)SO(4) was employed to modify Geotrichum sp. lipase for catalyzing biodiesel production from waste cooking oil. Compared to single modification of protein coating with K(2)SO(4), the dual modification of
Jingjing Zheng et al.
Bioresource technology, 110, 224-231 (2012-02-15)
This study investigated the preparation and characteristics of protein-coated microcrystals (PCMCs) from Pseudomonas cepacia lipase (PS) and K(2)SO(4), and their application in biodiesel synthesis, via single factorial experiments and response surface methodology (RSM), the optimized PCMC-PS exhibited high activity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service