Skip to Content
Merck
All Photos(1)

Key Documents

P4963

Millipore

Peptone Primatone® RL

suitable for microbiology

Synonym(s):

Peptone from animal tissue, Peptone from meat, Peptone from Animal tissue

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

biological source

animal

Quality Level

form

powder

shelf life

Limited shelf life, expiry date on the label

packaging

pkg of 100 g

storage condition

(Tightly closed)

loss

≤11% loss on drying

pH

6.9-7.6

application(s)

food and beverages
microbiology

SMILES string

O(C)C(=O)C(CCC(CC)(CC)C)C(=O)OC

InChI

1S/C13H24O4/c1-6-13(3,7-2)9-8-10(11(14)16-4)12(15)17-5/h10H,6-9H2,1-5H3

InChI key

AIUDWMLXCFRVDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Peptone Primatone® RL is widely used in industrial microbiology as a nutrient source for the growth of microorganisms in large-scale fermentation processes. Peptone Primatone® RL, also known as peptic meat peptone, is a water-soluble protein hydrolysate obtained from the pancreatic digestion of meat and used as a nutrient source in microbial culture media. It provides a wide range of nutrients, including amino acids, vitamins, minerals, and peptides, which support the growth of diverse microorganisms. It is particularly useful for the cultivation of fastidious and heterotrophic microbes that require complex nutrient sources.

Application

Peptone Primatone® RL is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts, and molds from various pharmaceutical, environmental, and food and beverage samples.

Legal Information

Primatone is a registered trademark of Kerry Group

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyadharshini Ramachandran et al.
Journal of microbiology and biotechnology, 23(3), 351-356 (2013-03-07)
A high beta-glucosidase (BGL)-producing strain, Stereum hirsutum, was identified and isolated and showed a maximum BGL activity (10.4 U/ml) when cultured with Avicel and tryptone as the carbon and nitrogen sources, respectively. In comparison with other BGLs, BGL obtained from
Takashi Kuda et al.
Food chemistry, 134(4), 1719-1723 (2013-02-28)
Listeria monocytogenes causes listeriosis in humans, mainly through the consumption of ready-to-eat foods such as cheese. Immunocompromised persons, the elderly, and pregnant women and their fetuses or newborns are at the highest risk for the infection. We examined the effects
Neeraj Bala et al.
Journal of basic microbiology, 52(4), 383-389 (2011-11-05)
The incubation of whole Bacillus alcalophilus cells grown on a mineral supplemented medium (MSM) containing 1% (w/v) sucrose as carbon source, 1.2% (w/v) tryptone as nitrogen source at pH 6.5 and temperature 30 °C in 24 h kinetically resolved benzyl
Aiqi Fang et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(32), 13225-13229 (2009-08-05)
In the hope of developing a vaccine against Clostridium difficile based on its toxin(s), we have developed a fermentation medium for the bacterium that results in the formation of Toxin A and contains no meat or dairy products, thus obviating
M Taskin et al.
Journal of applied microbiology, 111(4), 826-834 (2011-07-19)
Peptones are one of the most expensive constituents of microbial media. This study was undertaken to prepare the peptone from waste chicken feathers through a new process. The chemical analysis of chicken feather peptone (CFP) was performed. The ability of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service