Skip to Content
Merck
All Photos(1)

Key Documents

P2125

Supelco

D-Pantethine

analytical standard

Synonym(s):

Bis(N-pantothenylamidoethyl) disulfide, LBF factor, Pantethein oxidized form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H42N4O8S2
CAS Number:
Molecular Weight:
554.72
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic (organic)

Quality Level

grade

analytical standard

Assay

≥90%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

application(s)

forensics and toxicology
veterinary
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/C22H42N4O8S2/c1-21(2,13-27)17(31)19(33)25-7-5-15(29)23-9-11-35-36-12-10-24-16(30)6-8-26-20(34)18(32)22(3,4)14-28/h17-18,27-28,31-32H,5-14H2,1-4H3,(H,23,29)(H,24,30)(H,25,33)(H,26,34)/t17-,18-/m0/s1

InChI key

DJWYOLJPSHDSAL-ROUUACIJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

H Matsushima et al.
Experimental eye research, 64(3), 387-395 (1997-03-01)
This study of lens protein composition found that some cytoskeletal proteins were degraded during the earliest stages of cataract formation. Cataract was induced in 13-14 day old rats by a single subcutaneous injection of sodium selenite (19 mumol kg-1). By
J J Harding
Drugs & aging, 18(7), 473-486 (2001-08-03)
Cataract is the major cause of blindness and of visual impairment worldwide, so its prevention is of the greatest importance. At present no drug therapy is licensed for use in the UK or the US, so the only treatment for
M F McCarty
Medical hypotheses, 56(3), 314-317 (2001-05-22)
Pantethine is a versatile and well-tolerated hypolipidemic agent whose efficacy in this regard appears to be mediated by its catabolic product cystamine, a nucleophile which avidly attacks disulfide groups. An overview of pantethine research suggests that the hypotriglyceridemic activity of
John A Rumberger et al.
Nutrition research (New York, N.Y.), 31(8), 608-615 (2011-09-20)
Safety and efficacy of a biologically active derivative of vitamin B(5) (pantethine) on total cholesterol (TC) and low-density lipoprotein cholesterol (LDL-C) metabolism was studied in North American subjects at conventional low to moderate cardiovascular disease (CVD) risk. A total of
M J Webster
European journal of applied physiology and occupational physiology, 77(6), 486-491 (1998-07-03)
The purpose of this study was to investigate the physiological and performance responses to supplementation with allithiamin and pantethine. On two separate occasions, six highly trained cyclists [maximum O2 consumption or VO2max 61.8 (2.1) ml x kg(-1) x min(-1)] performed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service