Skip to Content
Merck
All Photos(2)

Key Documents

861251

Sigma-Aldrich

Neutral Red

certified by the Biological Stain Commission

Synonym(s):

3-Amino-7-dimethylamino-2-methylphenazine hydrochloride, Basic Red 5, Toluylene red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78
Colour Index Number:
50040
Beilstein:
3918740
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, 50-60%

technique(s)

titration: suitable

pH

6.8-8.0, red to yellow

mp

290 °C (dec.) (lit.)

solubility

water: 1 mg/mL

λmax

540 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.CN(C)c1ccc2nc3cc(C)c(N)cc3nc2c1

InChI

1S/C15H16N4.ClH/c1-9-6-13-15(8-11(9)16)18-14-7-10(19(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H

InChI key

PGSADBUBUOPOJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Neutral red has been used as a staining agent for micro- and meso-plankton. It has been used as a staining agent for nematocytes.
Useful as an indicator for preparing neutral red paper, and as a biological stain.

Biochem/physiol Actions

Neutral red is useful as an indicator for preparing neutral red paper, and as a biological stain. Neutral dye has low toxicity and is a vital dye. It can also be used as a pH indicator. It is used for cell viability assay. The dye accumulates in the lysosomes of living healthy cells, but not in damaged cells.

Suitability

Certified for use in the method of Doan & Ralph for the supravital staining of living blood cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dead or alive? Viability assessment of micro-and mesoplankton.
Zetsche EV and Meysman FJ
Journal of Plankton Research, 34, 493-509 (2012)
Cell Growth, Differentiaton and Senescence (1999)
The Anatomical Record, 27, 241-241 (1924)
Clung,C.E., Ed.
Handbook of Microscopical Technique, 571-571 (1950)
PH and Brain Function (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service