Skip to Content
Merck
All Photos(4)

Key Documents

M9929

Sigma-Aldrich

Methotrexate

meets USP testing specifications

Synonym(s):

Methotrexate hydrate, 4-Amino-10-methylfolic acid hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, L-Amethopterin hydrate, Antifolan hydrate, MTX hydrate, Methylaminopterin hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N8O5 · xH2O
CAS Number:
Molecular Weight:
454.44 (anhydrous basis)
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

98.0-102.0%

form

powder

storage condition

(Tightly closed. Dry. Keep in a well-ventilated place. )

technique(s)

activity assay: suitable

impurities

≤0.1% Residue on ignition (Ash)
≤12.0% water (Karl Fischer)

color

yellow to orange-yellow

mp

185-204 °C (365 - 399 °F)

solubility

water: insoluble

antibiotic activity spectrum

parasites

application(s)

metabolomics
microbiology

Mode of action

DNA synthesis

storage temp.

−20°C

SMILES string

[H]O[H].CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C20H22N8O5.H2O/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30;/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27);1H2/t13-;/m0./s1

InChI key

FPJYMUQSRFJSEW-ZOWNYOTGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems.
Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems. Also effective in treatment of pyrimethamine-resistant Plasmodium vivax malaria parasites.
Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems. Also shows immunosuppressive effects in, e.g., rheumatoid arthritis.

Biochem/physiol Actions

Methotrexate is an allosteric inhibititor of dihydrofolate reductase (DHFR), the enzyme that catalyzes the conversion of dihydrofolate to tetrahydrofolate. Since tetrahydrolfolate is required for purine and pyrimidine synthesis, methotrexate treatment results in the inhibition of DNA and RNA synthesis.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

related product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hany Elsawy et al.
Bioscience reports, 40(6) (2020-05-28)
Methotrexate (MTX) is an efficient chemotherapeutic and immunosuppressant drug, but the hepatotoxicity of MTX limits its clinical use. Naringin (Nar) is a flavonoid derived from Citrus paradise, and has been shown to possess several pharmacological activities, including free-radical scavenging and
Asma Achek et al.
EBioMedicine, 52, 102645-102645 (2020-02-06)
TLRs are some of the actively pursued drug-targets in immune disorders. Owing to a recent surge in the cognizance of TLR structural biology and signalling pathways, numerous therapeutic modulators, ranging from low-molecular-weight organic compounds to polypeptides and nucleic acid agents
Acharya Balkrishna et al.
Scientific reports, 9(1), 8025-8025 (2019-05-31)
Rheumatoid arthritis (RA) is a chronic inflammatory autoimmune disorder that affects joints of hands and feet and introduces injury in secondary organs such as cardiac tissue. In the present study, we induced RA in male Balb/c mice (CAIA) using collagen-antibody
Shih-Chao Lin et al.
Plants (Basel, Switzerland), 9(11) (2020-11-14)
Crotonoside, a guanosine analog originally isolated from Croton tiglium, is reported to be a potent tyrosine kinase inhibitor with immunosuppressive effects on immune cells. Due to its potential immunotherapeutic effects, we aimed to evaluate the anti-arthritic activity of crotonoside and
Jun-Young Park et al.
International journal of nanomedicine, 16, 4943-4957 (2021-07-31)
Understanding structural interactions between the active drug and conjugated nanoparticles is critical for optimizing intracellular drug transport and for increasing nano drug efficacy. In this regard, analyzing the conformational deformation of conjugated drugs surrounding nanoparticles is essential to understand the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service