Skip to Content
Merck
All Photos(1)

Key Documents

M0300000

Menadione

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Methyl-1,4-naphthoquinone, Vitamin K3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O2
CAS Number:
Molecular Weight:
172.18
Beilstein:
1908453
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

menadione

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

105-107 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=CC(=O)c2ccccc2C1=O

InChI

1S/C11H8O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6H,1H3

InChI key

MJVAVZPDRWSRRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Menadione, a fat-soluble vitamin, is an antihemorrhagic compound that plays an important role in blood coagulation. It takes part in blood clotting by activating prothrombin, the thrombin precursor.

Application

Menadione may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by chromatography and spectrophotometry techniques.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Menadione
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 13(2), 2998-2998 (2008)
Spectrophotometric methods for the rapid determination of menadione and menadione sodium bisulphite and their application in pharmaceutical preparations
Nagaraja P, et al.
Journal of Pharmaceutical and Biomedical Analysis, 28(1), 161-168 (2002)
Spectrophotometric determination of menadione and menadione sodium bisulfite in pharmaceutical preparations
Sastry CSP, et al.
International Journal of Pharmaceutics, 39(1-2), 137-140 (1987)
GC-FID and HPLC-DAD methods for the determination of menadione sodium bisulphite directly and by converting menadione sodium bisulphite to menadione in pharmaceutical preparation
Demirkaya-Miloglu F, et al.
The Iranian Journal of Pharmaceutical Research, 13(2), 353-353 (2014)
Y Usui et al.
The American journal of clinical nutrition, 51(5), 846-852 (1990-05-01)
Vitamin K deficiency has been reported in patients who were treated with antibiotics and placed on poor diets after surgery. High-performance liquid chromatography (HPLC) was used to study the influence of dietary intake on vitamin K concentrations in surgical patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service