Skip to Content
Merck
All Photos(1)

Documents

H1400000

Hydrocortisone acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hydrocortisone 21-acetate, 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate, 17-α-Hydroxycorticosterone acetate, 17-Hydroxycorticosterone 21-acetate, 21-Acetoxy-4-pregnene-11β,17α-diol-3,20-dione, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O6
CAS Number:
Molecular Weight:
404.50
Beilstein:
2066841
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hydrocortisone

manufacturer/tradename

EDQM

mp

223 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-18,20,26,28H,4-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1

InChI key

ALEXXDVDDISNDU-JZYPGELDSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydrocortisone acetate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biljana Jancic-Stojanović et al.
Journal of AOAC International, 93(1), 102-107 (2010-03-26)
An RP-HPLC method has been optimized and validated for the simultaneous determination of hydrocortisone acetate and of lidocaine in suppositories. For the method optimization, response surface methodology was applied, and the obtained model was tested using analysis of variance. The
Jaleh Varshosaz et al.
International journal of colorectal disease, 25(10), 1159-1165 (2010-07-30)
Anti-inflammatory drugs with high potency and low systemic adverse effects, such as budesonide, are drugs of choice for the treatment of ulcerative colitis (UC). Budesonide controlled-release formulations are now being used to induce and maintain clinical remission of Crohn's disease.
B Ramírez Andrade et al.
Journal of dairy science, 91(6), 2299-2306 (2008-05-20)
Lactation artificially induced (ART) by steroid hormones and natural lactation (NAT) after lambing were compared in 2 dairy sheep breeds (Manchega and Lacaune) in 2 experiments conducted during winter and spring. In experiment 1, ART ewes (14 Manchega and 9
Richard D Walton et al.
Heart rhythm, 7(12), 1843-1849 (2010-09-08)
Epifluorescence imaging using voltage-sensitive dyes has provided unique insights into cardiac electrical activity and arrhythmias. However, conventional dyes use blue-green excitation light, which has limited depth penetration. The aim of this study was to demonstrate that combining a short and
Shu Li et al.
Journal of mass spectrometry : JMS, 47(8), 1074-1082 (2012-08-18)
The applicability of liquid chromatography-mass spectrometry (LC/MS) is often limited by throughput. The sharing of a mass spectrometer with multiple LCs significantly improves throughput; however, the reported systems have not been designed to fully utilize the MS duty cycle, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service