Skip to Content
Merck
All Photos(1)

Key Documents

G3285

Supelco

D-(+)-Glucose solution

1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1281608
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

grade

analytical standard

Quality Level

analyte chemical class(es)

sugars (glucose)

concentration

1 mg/mL in 0.1% benzoic acid

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Component of Glucose (GO) Assay Kit (GAGO-20).

Analysis Note

Traceable to a NIST standard

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Rezaul Islam Khan et al.
Bio-protocol, 8(9), e2826-e2826 (2018-05-05)
Cyanobacteria, which have the extraordinary ability to grow using sunlight and carbon dioxide, are emerging as a green host to produce value-added products. Exploitation of this highly promising host to make products may depend on the ability to modulate the
Felix B Meyer et al.
BMC cancer, 21(1), 629-629 (2021-05-29)
Despite considerable medical proceedings, cancer is still a leading cause of death. Major problems for tumor therapy are chemoresistance as well as toxic side effects. In recent years, the additional treatment with the antidiabetic drug metformin during chemotherapy showed promising
Maria Paula Zappia et al.
eLife, 10 (2021-07-13)
The E2F transcription factors play a critical role in controlling cell fate. In Drosophila, the inactivation of E2F in either muscle or fat body results in lethality, suggesting an essential function for E2F in these tissues. However, the cellular and
Romana Stark et al.
Biochimica et biophysica acta, 1840(4), 1313-1330 (2013-11-02)
Plasma glucose levels are tightly regulated within a narrow physiologic range. Insulin-mediated glucose uptake by tissues must be balanced by the appearance of glucose from nutritional sources, glycogen stores, or gluconeogenesis. In this regard, a common pathway regulating both glucose
Yu-ting Alex Chiang et al.
American journal of physiology. Endocrinology and metabolism, 306(7), E707-E722 (2013-12-26)
p21-Activated protein kinases (PAKs) are centrally involved in a plethora of cellular processes and functions. Their function as effectors of small GTPases Rac1 and Cdc42 has been extensively studied during the past two decades, particularly in the realms of cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service