Skip to Content
Merck
All Photos(1)

Key Documents

F0041000

Fenbufen

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

γ-Oxo-(1,1′-biphenyl)-4-butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CO(CH2)2CO2H
CAS Number:
Molecular Weight:
254.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fenbufen

manufacturer/tradename

EDQM

mp

184-187 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CCC(=O)c1ccc(cc1)-c2ccccc2

InChI

1S/C16H14O3/c17-15(10-11-16(18)19)14-8-6-13(7-9-14)12-4-2-1-3-5-12/h1-9H,10-11H2,(H,18,19)

InChI key

ZPAKPRAICRBAOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fenbufen EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Kerwar
The American journal of medicine, 75(4B), 62-69 (1983-10-31)
Fenbufen (Cinopal) is an orally active nonsteroidal anti-inflammatory drug with analgesic and antipyretic activity. Like clinically used nonsteroidal anti-inflammatory drugs, it shows activity in a wide spectrum of laboratory tests in mice, rats, guinea pigs, and dogs. Fenbufen has a
A E Sloboda et al.
Arzneimittel-Forschung, 30(4A), 716-721 (1980-01-01)
gamma-Oxo(1,1'-biphenyl)-4-butanoic acid (fenbufen) was shown to be an orally and parenterally effective nonsteroidal antiinflammatory, analgesic and antipyretic agent in a variety of animal species. Like other clinically active antiinflammatory drugs such as acetylsalicylic acid (ASA), indometacin and phenylbutazone, fenbufen has
R Andrew Moore et al.
The Cochrane database of systematic reviews, (4)(4), CD007547-CD007547 (2009-10-13)
Fenbufen is a non-selective non-steroidal anti-inflammatory drug (NSAID), used to treat acute and chronic painful conditions. There is no known systematic review of its use in acute postoperative pain. To assess efficacy, duration of action, and associated adverse events of
C Kohler et al.
Arzneimittel-Forschung, 30(4A), 702-707 (1980-01-01)
gamma-Oxo(1,1'-biphenyl)-4-butanoic acid (fenbufen) and biphenylacetic acid have effects on platelets similar to other non-steroidal antiinflammatory agents. In vitro biphenylacetic acid (BPAA), a metabolite of fenbufen, is more potent than fenbufen and in vivo metabolism of fenbufen to BPAA is probably
R N Brogden et al.
Drugs, 21(1), 1-22 (1981-01-01)
Fenbufen is a phenylalkanoic acid derivative with analgesic and anti-inflammatory activity. The anti-inflammatory activity appears to reside in the metabolites. Published data indicate that fenbufen 600 to 1000mg daily is comparable in effectiveness to therapeutic doses (3 to 4g) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service