Skip to Content
Merck
All Photos(3)

Key Documents

D9680

Sigma-Aldrich

1,4-Dithioerythritol

BioXtra, ≥99.0%

Synonym(s):

erythro-1,4-Dimercapto-2,3-butanediol, erythro-2,3-Dihydroxy-1,4-butanedithiol, Cleland’s reagent, DTE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719756
EC Number:
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
NACRES:
NA.31

product line

BioXtra

Quality Level

Assay

≥99.0%

form

powder

reaction suitability

reagent type: reductant

impurities

Insoluble matter, passes filter test
≤0.0005% Phosphorus (P)

pH

4.5-6.5 (0.1 M in H2O)

mp

82-84 °C (lit.)

solubility

H2O: 0.1 M, clear, colorless

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.05%
Mg: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.001%

absorption

≤0.4 at 280 in H2O at 0.1 M
≤0.6 at 260 in H2O at 0.1 M

storage temp.

2-8°C

SMILES string

O[C@@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4+

InChI key

VHJLVAABSRFDPM-ZXZARUISSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for maintaining −SH groups in the reduced state; quantitatively reduces disulfides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gry Freja Skovsted et al.
Nutrients, 9(7) (2017-07-04)
Vitamin C (vitC) deficiency is associated with increased cardiovascular disease risk, but its specific interplay with arteriolar function is unclear. This study investigates the effect of vitC deficiency in guinea pigs on plasma biopterin status and the vasomotor responses in
Sam John et al.
Current protocols in molecular biology, Chapter 27, Unit 21-Unit 21 (2013-07-04)
DNase I-seq is a global and high-resolution method that uses the nonspecific endonuclease DNase I to map chromatin accessibility. These accessible regions, designated as DNase I hypersensitive sites (DHSs), define the regulatory features, (e.g., promoters, enhancers, insulators, and locus control
András Kolozsi et al.
Inorganic chemistry, 47(9), 3832-3840 (2008-04-03)
The aqueous solutions of arsenous acid with the meso and racemic forms of 1,4-dithiol-butane-2,3-diol, namely, dithioerythritol (dte) and dithiothreitol (dtt), respectively, were titrated pH-metrically in different molar ratios. The p K a values determined for As(OH) 3, and dtt were
Tanju Ceyhan et al.
Dalton transactions (Cambridge, England : 2003), 39(41), 9801-9814 (2010-09-18)
The phthalodinitrile derivative 3 was prepared by the reaction of 1,4-dithioerythritol 1 and 4-nitrophthalonitrile 2 in dry DMF as the solvent in the presence of K(2)CO(3) as the base by the method of nucleophilic substitution of an activated nitro group
Kenan Coyan et al.
Research in veterinary science, 89(3), 426-431 (2010-04-21)
The aim of this study was to investigate the effects of methionine and dithioerythritol, added to the Tris extender, on ram sperm motility and LPO (lipid peroxidation) and antioxidant capacities during liquid storage up to 72 h at 5°C. Ejaculates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service