Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,4-Dithioerythritol
Synonym(s):
erythro
-1,4-Dimercapto-2,3-butanediol,
erythro
-2,3-Dihydroxy-1,4-butanedithiol, Cleland’s reagent, DTE, 1,4-Dithioerythritol
Linear Formula:
HSCH
2
CH(OH)CH(OH)CH
2
SH
CAS Number:
6892-68-8
Molecular Weight:
154.25
EC Number:
229-998-8
Products
Brand
Millipore (1)
Sigma-Aldrich (4)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
D8161
1,4-Dithioerythritol,
BioReagent, for molecular biology, ≥99.0%
Expand
View Pricing
43794
1,4-Dithioerythritol,
≥99.0% (RT), BioUltra
Expand
View Pricing
D8255
1,4-Dithioerythritol,
≥99.0%
Expand
View Pricing
1.24511
1,4-Dithioerythritol,
for biochemistry
Expand
View Pricing
D9680
1,4-Dithioerythritol,
BioXtra, ≥99.0%
Expand
View Pricing